Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01308100_g1
          See other CHKB GE Assays ›
          Gene Symbol:
          Chkb ChkbCpt1b
          Gene Name:
          choline kinase beta
          Chkb-Cpt1b readthrough transcript (NMD candidate)
          Gene Aliases:
          Chetk, Chkl, Ck, Ck/Ek, Ck/Ek-beta, Ckb, Ckekb, Ek
          BC090627, Chkb-Cpt1b, Chkbcpt1b
          Chromosome Location:
          Chr.15: 89426349 - 89429927 on Build GRCm38
          Chromosome Location:
          Chr.15: 89416405 - 89429927 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm01308100_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chkb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007692.6 NP_031718.1 5 - 6 1006 84
          NR_037153.1 4 - 5 1175 84
          NR_037154.1 5 - 6 1040 84
          GenBank mRNA AB011000.1 - 5 - 6 978 84
          AB011001.1 - 3 - 4 1328 84
          AK078101.1 - 5 - 6 1000 84
          AK164379.1 - 2 - 3 1557 84
          AK170077.1 - 4 - 5 687 84
          BC068195.1 - 5 - 6 524 4164116 84
          BC069925.1 - 3 - 4 1327 84
          Gene Symbol: ChkbCpt1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_004843.2 4 - 5 1175 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002351.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chkb
          Click here to find siRNAs for Gene ChkbCpt1b

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          phosphatidylethanolamine biosynthetic process
          phosphatidylcholine biosynthetic process
          CDP-choline pathway
          muscle organ development
          phospholipid biosynthetic process
          phosphorylation
          nucleotide binding
          choline kinase activity
          ethanolamine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500016565.mm.1 TC1500002351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500016565.mm.1 NM_007692.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016565.mm.1 NM_007692.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016565.mm.1 NR_037153.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016565.mm.1 NR_037153.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016565.mm.1 NR_037154.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016565.mm.1 NR_037154.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016567.mm.1 TC1500002351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500016567.mm.1 NM_007692.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016567.mm.1 NM_007692.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016567.mm.1 NR_037153.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016567.mm.1 NR_037153.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016567.mm.1 NR_037154.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016567.mm.1 NR_037154.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016569.mm.1 TC1500002351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500016569.mm.1 NM_007692.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016569.mm.1 NM_007692.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016569.mm.1 NR_037153.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016569.mm.1 NR_037153.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016569.mm.1 NR_037154.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016569.mm.1 NR_037154.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016566.mm.1 TC1500002351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500016566.mm.1 NM_007692.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016566.mm.1 NM_007692.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016566.mm.1 NR_037153.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016566.mm.1 NR_037153.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016566.mm.1 NR_037154.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016566.mm.1 NR_037154.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016563.mm.1 TC1500002351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500016563.mm.1 NM_007692.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016563.mm.1 NM_007692.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016563.mm.1 NR_037153.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016563.mm.1 NR_037153.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016563.mm.1 NR_037154.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500016563.mm.1 NR_037154.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0