Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01309169_g1
          See other THOC6 GE Assays ›
          Gene Symbol:
          Thoc6
          Gene Name:
          THO complex 6 homolog (Drosophila)
          Gene Aliases:
          F830014G06Rik, Wdr58
          Chromosome Location:
          Chr.17: 23668618 - 23673903 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01309169_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thoc6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008425.1 NP_001008425.1 12 - 13 1162 68
          GenBank mRNA AK089746.1 - 9 - 10 1490 68
          BC085131.1 - 12 - 13 1162 5715435 68
          BC089551.1 - 13 - 14 1162 30303151 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001597.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thoc6

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          mRNA export from nucleus
          transport
          apoptotic process
          multicellular organism development
          central nervous system development
          RNA splicing
          negative regulation of apoptotic process
          viral mRNA export from host cell nucleus
          mRNA transport
          RNA binding
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092956 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169849 BC089551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169849 NM_001008425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169849 AK089746.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2169849 BC085131.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 BC089551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 NM_001008425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 AK089746.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 BC085131.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 BC089551.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 NM_001008425.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 AK089746.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P631568 BC085131.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485377 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157241 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110491 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169849 BC089551.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169849 NM_001008425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169849 AK089746.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2169849 BC085131.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 BC089551.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 NM_001008425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 AK089746.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 BC085131.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 BC089551.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 NM_001008425.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 AK089746.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P631568 BC085131.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092961 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03292144 - -
          Whole Mouse Genome G4122A A_52_P631568 BC089551.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631568 NM_001008425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631568 AK089746.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631568 BC085131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631568 BC089551.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631568 NM_001008425.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631568 AK089746.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P631568 BC085131.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111530 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113437 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13214726 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13214687 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13214711 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13214697 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31093029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092969 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448305 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448306 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448304 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448302 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448303 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448296 - -
          Affymetrix HT MG-430 PM Array Plate 1434827_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442321 - -
          Affymetrix Mouse Gene 1.1 ST Array 10448292 - -
          Mouse Genome 430 2.0 Array 1434827_PM_at - -
          Mouse Genome 430 2.0 Array 1434827_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 386612_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 386612_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700013932.mm.1 TC1700001597.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700013932.mm.1 NM_001008425.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013932.mm.1 NM_001008425.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013933.mm.1 TC1700001597.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700013933.mm.1 NM_001008425.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700013933.mm.1 NM_001008425.1 Refseq
          Mouse Expression Set 430 1434827_PM_at - -
          Mouse Expression Set 430 1434827_at - -
          Mouse Genome 430A 2.0 Array 1434827_PM_at - -
          Mouse Genome 430A 2.0 Array 1434827_at - -
          Affymetrix HT Mouse Genome 430A Array 1434827_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434827_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 693022 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100780600 - -
          Illumina mouse-6 v1.0 expression beadchip 100780600 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249706 - -
          Illumina Sentrix mouse-6.v1 bead array 100780600 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100780600 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2778428 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2778428 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0