Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01309718_g1
          See other DHRSX GE Assays ›
          Gene Symbol:
          Dhrsx
          Gene Name:
          dehydrogenase/reductase (SDR family) X chromosome
          Gene Aliases:
          C80877, D4Ertd296e, Dhrsxy, Pscad
          Chromosome Location:
          Chr.4|NT_166291: 15800 - 36213 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01309718_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dhrsx
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033326.2 NP_001028498.2 5 - 6 1010 84
          GenBank mRNA AJ296079.1 - 5 - 6 84
          AK157461.1 - 5 - 6 694 84
          BC046917.1 - 5 - 6 733 84
          BC138597.1 - 5 - 6 651 8861617 84
          BC138599.1 - 5 - 6 651 8861619 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC4_GL456216_random00000001.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dhrsx

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          metabolic process
          molecular_function
          oxidoreductase activity
          pinocarveol dehydrogenase activity
          chloral hydrate dehydrogenase activity
          hydroxymethylmethylsilanediol oxidase activity
          1-phenylethanol dehydrogenase activity
          myrtenol dehydrogenase activity
          cis-1,2-dihydroxy-1,2-dihydro-8-carboxynaphthalene dehydrogenase activity
          3-hydroxy-4-methyloctanoyl-CoA dehydrogenase activity
          2-hydroxy-4-isopropenylcyclohexane-1-carboxyl-CoA dehydrogenase activity
          cis-9,10-dihydroanthracene-9,10-diol dehydrogenase activity
          citronellol dehydrogenase activity
          naphthyl-2-hydroxymethyl-succinyl-CoA dehydrogenase activity
          2,4,4-trimethyl-1-pentanol dehydrogenase activity
          2,4,4-trimethyl-3-hydroxypentanoyl-CoA dehydrogenase activity
          1-hydroxy-4,4-dimethylpentan-3-one dehydrogenase activity
          endosulfan diol dehydrogenase activity
          endosulfan hydroxyether dehydrogenase activity
          3-hydroxy-2-methylhexanoyl-CoA dehydrogenase activity
          3-hydroxy-2,6-dimethyl-5-methylene-heptanoyl-CoA dehydrogenase activity
          versicolorin reductase activity
          ketoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644534 AK157461.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644534 BF714825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644534 NM_001033326.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644534 AK030649.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644534 AK157461.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644534 BF714825.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644534 NM_001033326.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P644534 AK030649.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644534 AK157461.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644534 BF714825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644534 NM_001033326.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644534 AK030649.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644534 AK157461.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644534 BF714825.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644534 NM_001033326.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P644534 AK030649.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P102867 AK157461.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P102867 BC138597.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P102867 BC138599.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P102867 AJ296079.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644534 AK157461.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644534 BF714825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644534 NM_001033326.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644534 AK030649.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644534 AK157461.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644534 BF714825.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644534 NM_001033326.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P644534 AK030649.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10598116 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598115 - -
          Affymetrix HT MG-430 PM Array Plate 1456484_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10598111 - -
          Mouse Genome 430 2.0 Array 1456484_PM_at - -
          Mouse Genome 430 2.0 Array 1456484_at - -
          Affymetrix Mouse Clariom S Array PSR4_GL456216_random00000009.mm.1 TC4_GL456216_random00000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR4_GL456216_random00000009.mm.1 NM_001033326.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_GL456216_random00000007.mm.1 TC4_GL456216_random00000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR4_GL456216_random00000007.mm.1 NM_001033326.2 Refseq
          Affymetrix Mouse Clariom S Array PSR4_GL456216_random00000008.mm.1 TC4_GL456216_random00000001.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR4_GL456216_random00000008.mm.1 NM_001033326.2 Refseq
          Mouse Expression Set 430 1456484_at - -
          Mouse Expression Set 430 1456484_PM_at - -
          Murine Genome U74v2 Set 93238_at - -
          Murine Genome U74v2 Set 93238_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456484_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456484_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 481425 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102230338 - -
          MouseWG-6 v2 Expression BeadChip 103440168 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221451 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2436891 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2436890 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2897020 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2897020 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0