Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01309897_m1
          See other NOS2 GE Assays ›
          Gene Symbol:
          Nos2
          Gene Name:
          nitric oxide synthase 2, inducible
          Gene Aliases:
          NOS-II, Nos-2, Nos2a, i-NOS, iNOS
          Chromosome Location:
          Chr.11: 78920859 - 78960232 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01309897_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nos2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010927.3 NP_035057.1 25 - 26 3325 77
          GenBank mRNA AF065919.1 - 25 - 26 3322 77
          AF065920.1 - 25 - 26 3322 77
          AF065921.2 - 25 - 26 3322 77
          AF065922.2 - 24 - 25 3195 77
          AF065923.2 - 24 - 25 3195 77
          AY090567.1 - 24 - 25 3145 77
          BC062378.1 - 25 - 26 3334 4978648 77
          M84373.1 - 25 - 26 3326 77
          M87039.1 - 25 - 26 3397 77
          M92649.1 - 25 - 26 3333 77
          U43428.1 - 25 - 26 3213 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001171.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nos2

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          ovulation from ovarian follicle
          response to hypoxia
          arginine catabolic process
          superoxide metabolic process
          nitric oxide biosynthetic process
          inflammatory response
          nitric oxide mediated signal transduction
          negative regulation of gene expression
          peptidyl-cysteine S-nitrosylation
          positive regulation of guanylate cyclase activity
          response to lipopolysaccharide
          cellular response to drug
          regulation of cell proliferation
          negative regulation of protein catabolic process
          defense response to bacterium
          negative regulation of blood pressure
          regulation of protein kinase activity
          positive regulation of vasodilation
          regulation of insulin secretion
          positive regulation of killing of cells of other organism
          oxidation-reduction process
          cellular response to lipopolysaccharide
          cellular response to interferon-gamma
          actin binding
          NADPH-hemoprotein reductase activity
          nitric-oxide synthase activity
          receptor binding
          iron ion binding
          protein binding
          calmodulin binding
          beta-catenin binding
          cAMP-dependent protein kinase regulator activity
          FMN binding
          oxidoreductase activity
          protein kinase binding
          heme binding
          tetrahydrobiopterin binding
          arginine binding
          protein homodimerization activity
          cadherin binding
          metal ion binding
          flavin adenine dinucleotide binding
          nitric-oxide synthase binding
          Hsp90 protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100010939.mm.1 NM_010927.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010939.mm.1 TC1100001171.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0