Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01310122_m1
          See other XRCC6 GE Assays ›
          Gene Symbol:
          Xrcc6
          Gene Name:
          X-ray repair complementing defective repair in Chinese hamster cells 6
          Gene Aliases:
          70kDa, G22p1, Ku70
          Chromosome Location:
          Chr.15: 81987826 - 82040085 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01310122_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xrcc6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010247.2 NP_034377.2 10 - 11 1542 84
          GenBank mRNA AB010282.1 - 10 - 11 1483 84
          AF483500.1 - 9 - 10 1422 84
          AF483501.1 - 9 - 10 1422 84
          AK088530.1 - 4 - 5 492 84
          AK143570.1 - 10 - 11 1517 84
          AK146392.1 - 10 - 11 1461 84
          AK146394.1 - 10 - 11 1461 84
          AK146396.1 - 10 - 11 1509 84
          BC031422.1 - 10 - 11 1491 3669715 84
          BC051085.1 - 10 - 11 2720 84
          M38700.1 - 9 - 10 1384 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000736.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Xrcc6

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          DNA repair
          double-strand break repair
          double-strand break repair via nonhomologous end joining
          DNA recombination
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          cellular response to DNA damage stimulus
          metabolic process
          response to ionizing radiation
          V(D)J recombination
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          positive regulation of neurogenesis
          cellular hyperosmotic salinity response
          cellular response to X-ray
          nucleotide binding
          transcription cis-regulatory region binding
          DNA binding
          DNA helicase activity
          damaged DNA binding
          double-stranded DNA binding
          double-stranded telomeric DNA binding
          RNA binding
          catalytic activity
          helicase activity
          ATP binding
          protein C-terminus binding
          hydrolase activity
          lyase activity
          telomeric DNA binding
          5'-deoxyribose-5-phosphate lyase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30380963 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30380965 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30380962 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P108567 BC031422.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P108567 BC051085.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P108567 AK146392.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P108567 AK146394.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P108567 AK146396.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 473064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 341849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 812880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 376590 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956100 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 142490 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 301419 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887516 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 332891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140418 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 127997 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83077 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156080 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116437 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P108567 BC031422.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P108567 BC051085.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P108567 AK146392.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P108567 AK146394.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P108567 AK146396.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P486738 M38700.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30380980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30380965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30380971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30380967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30380973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30380979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30380974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30380964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30380977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30380969 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116437 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08436923 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08436983 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03083372 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03083381 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156080 - -
          Whole Mouse Genome G4122A A_51_P486738 M38700.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108567 BC031422.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108567 BC051085.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108567 AK146392.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108567 AK146394.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108567 AK146396.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120275 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116437 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156080 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12188691 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12188678 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12188684 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30380963 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30380965 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30380962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30380989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30381086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12369279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30381084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380988 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11927011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12062438 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12369278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30380989 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10425635 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425649 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425639 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425638 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425650 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425648 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10425643 - -
          Affymetrix HT MG-430 PM Array Plate 1417437_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428504_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10425632 - -
          Mouse Genome 430 2.0 Array 1442250_at - -
          Mouse Genome 430 2.0 Array 1417437_PM_at - -
          Mouse Genome 430 2.0 Array 1428504_at - -
          Mouse Genome 430 2.0 Array 1428504_PM_at - -
          Mouse Genome 430 2.0 Array 1442250_PM_at - -
          Mouse Genome 430 2.0 Array 1417437_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14375_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14375_at - -
          Affymetrix Mouse Clariom S Array PSR1500005775.mm.1 TC1500000736.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500005775.mm.1 NM_010247.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500005775.mm.1 NM_010247.2 Refseq
          Mouse Expression Set 430 1417437_at - -
          Mouse Expression Set 430 1417437_PM_at - -
          Mouse Expression Set 430 1428504_at - -
          Mouse Expression Set 430 1428504_PM_at - -
          Murine Genome U74v2 Set 103036_PM_at - -
          Murine Genome U74v2 Set 103036_at - -
          Mouse Genome 430A 2.0 Array 1417437_at - -
          Mouse Genome 430A 2.0 Array 1417437_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417437_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417437_at - -
          Affymetrix HT Mouse Genome 430B Array 1428504_at - -
          Affymetrix HT Mouse Genome 430B Array 1428504_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2850450 - -
          Illumina mouse-6 v1.0 expression beadchip 3870528 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850450 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003870528 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216412 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1247814 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216412 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1247814 - -
          mouse-6 v1.1 (Illumina) scl014375.11_27-S - -
          mouse-6 v1.1 (Illumina) scl0002561.1_38-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850450 - -
          Illumina Sentrix mouse-6.v1 bead array 3870528 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002561.1_38-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl014375.11_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3870528 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850450 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1247814 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2862619 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216412 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216412 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1247814 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2862619 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl014375.11_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0002561.1_38-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0