Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01310713_m1
          See other LRRC63 GE Assays ›
          Gene Symbol:
          Lrrc63
          Gene Name:
          leucine rich repeat containing 63
          Gene Aliases:
          4921509B22Rik
          Chromosome Location:
          Chr.14: 75084303 - 75130962 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01310713_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lrrc63
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027581.1 NP_081857.1 9 - 10 1836 60
          GenBank mRNA AK014850.1 - 8 - 9 1568 60
          BC145796.1 - 9 - 10 1831 40131099 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002415.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P374521 BC145796.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P374521 NM_027581.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P374521 AK014850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P374521 BC145796.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P374521 NM_027581.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P374521 AK014850.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855570 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150112 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138119 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P374521 BC145796.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P374521 NM_027581.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P374521 AK014850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P374521 BC145796.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P374521 NM_027581.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P374521 AK014850.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150112 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08164891 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138119 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08164826 - -
          Whole Mouse Genome G4122A A_51_P374521 BC145796.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374521 NM_027581.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374521 AK014850.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374521 BC145796.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374521 NM_027581.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374521 AK014850.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150112 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138119 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08164891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29726002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11183447 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11698771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11362290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11695902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11183449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11698769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29725831 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10421614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10421613 - -
          Affymetrix HT MG-430 PM Array Plate 1429973_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10421605 - -
          Mouse Genome 430 2.0 Array 1429973_PM_at - -
          Mouse Genome 430 2.0 Array 1429973_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70859_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70859_at - -
          Affymetrix Mouse Clariom S Array PSR1400018727.mm.1 TC1400002415.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400018727.mm.1 NM_027581.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018727.mm.1 NM_027581.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018728.mm.1 TC1400002415.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400018728.mm.1 NM_027581.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400018728.mm.1 NM_027581.1 Refseq
          Mouse Expression Set 430 1429973_at - -
          Mouse Expression Set 430 1429973_PM_at - -
          Murine Genome U74v2 Set 137236_f_at - -
          Murine Genome U74v2 Set 137236_PM_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1429973_at - -
          Affymetrix HT Mouse Genome 430B Array 1429973_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 394722 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2590387 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2590387 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0