Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01310771_g1
          See other CD34 GE Assays ›
          Gene Symbol:
          Cd34
          Gene Name:
          CD34 antigen
          Gene Aliases:
          AU040960
          Chromosome Location:
          Chr.1: 194938821 - 194961292 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01310771_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd34
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001111059.1 NP_001104529.1 5 - 6 888 84
          NM_133654.3 NP_598415.1 5 - 6 888 84
          GenBank mRNA AK044121.1 - 5 - 6 824 84
          AK053874.1 - 5 - 6 824 84
          AK089010.1 - 5 - 6 794 84
          BC006607.1 - 5 - 6 804 3966337 84
          BC066820.1 - 5 - 6 761 84
          BU058977.1 - 3 - 4 371 84
          S69293.1 - 5 - 6 766 84
          S69299.1 - 5 - 6 743 84
          S69301.1 - 5 - 6 743 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001882.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd34

          Gene Ontology Categories:

          Function(s) Process(es)

          tissue homeostasis
          endothelial cell proliferation
          cell adhesion
          regulation of blood pressure
          negative regulation of gene expression
          hemopoiesis
          negative regulation of blood coagulation
          negative regulation of interleukin-2 production
          negative regulation of tumor necrosis factor production
          positive regulation of interleukin-10 production
          paracrine signaling
          positive regulation of odontogenesis
          negative regulation of nitric oxide biosynthetic process
          positive regulation of angiogenesis
          leukocyte migration
          hematopoietic stem cell proliferation
          positive regulation of transforming growth factor beta production
          positive regulation of granulocyte colony-stimulating factor production
          extracellular exosome assembly
          cell-cell adhesion
          negative regulation of cellular response to heat
          negative regulation of cellular response to hypoxia
          positive regulation of glial cell-derived neurotrophic factor production
          negative regulation of neuron death
          positive regulation of vasculogenesis
          transcription factor binding
          carbohydrate binding
          sulfate binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100015638.mm.1 TC0100001882.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100015638.mm.1 NM_001111059.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015638.mm.1 NM_001111059.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015638.mm.1 NM_133654.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100015638.mm.1 NM_133654.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0