Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01310829_g1
          See other MFSD2B GE Assays ›
          Gene Symbol:
          Mfsd2b
          Gene Name:
          major facilitator superfamily domain containing 2B
          Gene Aliases:
          Gm1964
          Chromosome Location:
          Chr.12: 4862438 - 4875044 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01310829_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mfsd2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033488.2 NP_001028660.2 11 - 12 1169 58
          GenBank mRNA AK156138.1 - 12 - 13 1321 58
          AK172423.1 - 11 - 12 1170 58
          BC145584.1 - 11 - 12 1182 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001367.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202815 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202813 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202814 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159746 AK172423.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159746 AK162620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159746 AK171660.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159746 AK156138.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159746 AK157892.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159740 AK162620.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159740 AK171660.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2159740 NM_001033488.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852704 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546739 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 414746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 629973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789249 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886747 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 609359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34773 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164450 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159740 AK162620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159740 AK171660.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159740 NM_001033488.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159746 AK172423.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159746 AK162620.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159746 AK171660.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159746 AK156138.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2159746 AK157892.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28202810 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164450 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07524561 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164450 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202815 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202814 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28202813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28202749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10143694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09992970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10284850 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10231515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10231516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10143699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10117006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10143697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202704 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10143705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10143703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10369238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202707 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10332074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10348891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09817990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28202702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P10143704 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10399322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399325 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399321 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399329 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399323 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399328 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399326 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399327 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399324 - -
          Affymetrix Mouse Gene 1.0 ST Array 10399320 - -
          Affymetrix HT MG-430 PM Array Plate 1457933_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1442000_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10399314 - -
          Mouse Genome 430 2.0 Array 1442000_at - -
          Mouse Genome 430 2.0 Array 1457933_PM_at - -
          Mouse Genome 430 2.0 Array 1457933_at - -
          Mouse Genome 430 2.0 Array 1442000_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 432628_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 432628_at - -
          Affymetrix Mouse Clariom S Array PSR1200009801.mm.1 TC1200001367.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009801.mm.1 NM_001033488.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009801.mm.1 NM_001033488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009802.mm.1 TC1200001367.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009802.mm.1 NM_001033488.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009802.mm.1 NM_001033488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009800.mm.1 TC1200001367.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009800.mm.1 NM_001033488.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009800.mm.1 NM_001033488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009804.mm.1 TC1200001367.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009804.mm.1 NM_001033488.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009804.mm.1 NM_001033488.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009799.mm.1 TC1200001367.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200009799.mm.1 NM_001033488.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200009799.mm.1 NM_001033488.2 Refseq
          Mouse Expression Set 430 1457933_at - -
          Mouse Expression Set 430 1442000_at - -
          Mouse Expression Set 430 1457933_PM_at - -
          Mouse Expression Set 430 1442000_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1442000_at - -
          Affymetrix HT Mouse Genome 430B Array 1442000_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457933_at - -
          Affymetrix HT Mouse Genome 430B Array 1457933_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161168 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161168 - -
          mouse-6 v1.1 (Illumina) GI_85702024-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161168 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161168 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0