Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200009778.mm.1 |
TC1200001364.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009778.mm.1 |
NM_175431.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009778.mm.1 |
NM_175431.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009780.mm.1 |
TC1200001364.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009780.mm.1 |
NM_175431.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009780.mm.1 |
NM_175431.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009779.mm.1 |
TC1200001364.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009779.mm.1 |
NM_175431.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009779.mm.1 |
NM_175431.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009781.mm.1 |
TC1200001364.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200009781.mm.1 |
NM_175431.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200009781.mm.1 |
NM_175431.4 |
Refseq |