Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01311230_m1
          See other DDIAS GE Assays ›
          Gene Symbol:
          Ddias
          Gene Name:
          DNA damage-induced apoptosis suppressor
          Gene Aliases:
          4632434I11Rik, noxin
          Chromosome Location:
          Chr.7: 92857525 - 92874286 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01311230_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddias
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080995.1 NP_001074464.1 6 - 7 661 108
          GenBank mRNA AK014608.2 - 5 - 6 581 108
          AK154915.1 - 5 - 6 564 108
          BC066148.1 - 5 - 6 555 30086532 108
          DQ400346.1 - 5 - 6 595 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003743.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          cellular response to DNA damage stimulus
          cell cycle
          spermatid development
          hemopoiesis
          cellular response to UV
          regulation of cell cycle
          cellular response to cytokine stimulus
          cellular response to hydroperoxide
          cellular response to gamma radiation
          negative regulation of fibroblast apoptotic process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369252 AK014608.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369252 AK014608.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625023 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 160941 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874774 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 280208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 1652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144767 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168493 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155389 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369252 AK014608.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369252 AK014608.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06282435 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155389 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168493 - -
          Whole Mouse Genome G4122A A_51_P369252 AK014608.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369252 AK014608.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111212 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155389 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168493 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06031178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06031193 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06031171 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06031191 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06031190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25326986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25327064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25326974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25326969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06056589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25326968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06150896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06031188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06031169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06567663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25326980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06266007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25326977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25326975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327074 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06175484 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06031170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25326970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25326972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25326976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06128684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25327096 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10565574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565575 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565576 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565573 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565571 - -
          Affymetrix Mouse Gene 1.0 ST Array 10565572 - -
          Affymetrix HT MG-430 PM Array Plate 1434240_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429734_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10565570 - -
          Mouse Genome 430 2.0 Array 1434240_at - -
          Mouse Genome 430 2.0 Array 1429734_at - -
          Mouse Genome 430 2.0 Array 1434240_PM_at - -
          Mouse Genome 430 2.0 Array 1429734_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74041_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74041_at - -
          Affymetrix Mouse Clariom S Array PSR0700031555.mm.1 TC0700003743.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700031555.mm.1 NM_001080995.2 Refseq
          Mouse Expression Set 430 1429734_PM_at - -
          Mouse Expression Set 430 1434240_PM_at - -
          Mouse Expression Set 430 1434240_at - -
          Mouse Expression Set 430 1429734_at - -
          Murine Genome U74v2 Set 132418_at - -
          Murine Genome U74v2 Set 132418_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434240_at - -
          Affymetrix HT Mouse Genome 430B Array 1429734_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429734_at - -
          Affymetrix HT Mouse Genome 430B Array 1434240_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 817048 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2230451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2660829 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2660829 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2446902 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2660829 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix