Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400000673.mm.1 |
TC1400000075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000673.mm.1 |
NM_025938.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000673.mm.1 |
NM_025938.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000678.mm.1 |
TC1400000075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000678.mm.1 |
NM_025938.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000678.mm.1 |
NM_025938.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000677.mm.1 |
TC1400000075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000677.mm.1 |
NM_025938.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000677.mm.1 |
NM_025938.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000676.mm.1 |
TC1400000075.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400000676.mm.1 |
NM_025938.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400000676.mm.1 |
NM_025938.4 |
Refseq |