Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01311582_g1
          See other PXK GE Assays ›
          Gene Symbol:
          Pxk
          Gene Name:
          PX domain containing serine/threonine kinase
          Gene Aliases:
          C230080L11Rik, D14Ertd813e, MONaKA
          Chromosome Location:
          Chr.14: 8098191 - 8165117 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01311582_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pxk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145458.3 NP_663433.2 15 - 16 1496 84
          NM_178279.2 NP_840063.1 15 - 16 1496 84
          GenBank mRNA AK028501.1 - 15 - 16 1446 84
          AK048910.1 - 15 - 16 1456 84
          AK147037.1 - 15 - 16 1496 84
          AK150337.1 - 15 - 16 1481 84
          AK150413.1 - 15 - 16 1481 84
          AK150660.1 - 15 - 16 1497 84
          AK151098.1 - 15 - 16 1457 84
          AK152058.1 - 15 - 16 1455 84
          AK152789.1 - 15 - 16 1455 84
          AK153067.1 - 15 - 16 1455 84
          AK156146.1 - 15 - 16 1462 84
          AK157364.1 - 15 - 16 1490 84
          AK159541.1 - 3 - 4 229 84
          AK166036.1 - 15 - 16 1422 84
          AK169112.1 - 15 - 16 1456 84
          AK170361.1 - 15 - 16 1492 84
          AK172463.1 - 15 - 16 1498 84
          BC016131.1 - 15 - 16 1466 4922274 84
          DQ095196.1 - 15 - 16 1394 84
          DQ095197.1 - 15 - 16 1394 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000076.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pxk

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          inflammatory response
          negative regulation of ATPase activity
          negative regulation of ion transport
          actin binding
          protein kinase activity
          ATP binding
          protein C-terminus binding
          transferase activity, transferring phosphorus-containing groups
          phosphatidylinositol binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400000706.mm.1 TC1400000076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400000706.mm.1 NM_145458.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000706.mm.1 NM_145458.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000706.mm.1 NM_178279.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000706.mm.1 NM_178279.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000705.mm.1 TC1400000076.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400000705.mm.1 NM_145458.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000705.mm.1 NM_145458.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000705.mm.1 NM_178279.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400000705.mm.1 NM_178279.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0