Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01312744_g1
          See other ALDH1A3 GE Assays ›
          Gene Symbol:
          Aldh1a3
          Gene Name:
          aldehyde dehydrogenase family 1, subfamily A3
          Gene Aliases:
          ALDH6, RALDH3, V1
          Chromosome Location:
          Chr.7: 66390892 - 66427477 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01312744_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aldh1a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053080.3 NP_444310.3 4 - 5 541 57
          GenBank mRNA AF152359.1 - 4 - 5 496 57
          AF246711.1 - 4 - 5 489 57
          AF253409.1 - 4 - 5 528 57
          AF280404.1 - 4 - 5 493 57
          AK043530.1 - 4 - 5 538 57
          AK086764.1 - 4 - 5 554 57
          AK147006.1 - 4 - 5 542 57
          BC026667.1 - 3 - 4 288 57
          BC058277.1 - 4 - 5 485 6515355 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003437.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aldh1a3

          Gene Ontology Categories:

          Function(s) Process(es)

          optic cup morphogenesis involved in camera-type eye development
          retinoic acid biosynthetic process
          cellular aldehyde metabolic process
          locomotory behavior
          metabolic process
          nucleus accumbens development
          embryonic camera-type eye development
          inner ear morphogenesis
          retinoic acid metabolic process
          retinal metabolic process
          positive regulation of apoptotic process
          embryonic eye morphogenesis
          positive regulation of retinoic acid receptor signaling pathway
          neuromuscular process controlling balance
          righting reflex
          olfactory pit development
          face development
          retinal dehydrogenase activity
          3-chloroallyl aldehyde dehydrogenase activity
          aldehyde dehydrogenase (NAD+) activity
          aldehyde dehydrogenase [NAD(P)+] activity
          oxidoreductase activity
          oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
          protein homodimerization activity
          thyroid hormone binding
          NAD+ binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700029320.mm.1 TC0700003437.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029320.mm.1 NM_053080.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029320.mm.1 NM_053080.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029321.mm.1 TC0700003437.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029321.mm.1 NM_053080.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029321.mm.1 NM_053080.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline