Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400017793.mm.1 |
TC1400002301.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017793.mm.1 |
NM_007402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017793.mm.1 |
NM_007402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017794.mm.1 |
TC1400002301.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017794.mm.1 |
NM_007402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017794.mm.1 |
NM_007402.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017792.mm.1 |
TC1400002301.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400017792.mm.1 |
NM_007402.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400017792.mm.1 |
NM_007402.2 |
Refseq |