Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01315246_m1
          See other 1700108F19RIK GE Assays ›
          Gene Symbol:
          1700108F19Rik
          Gene Name:
          RIKEN cDNA 1700108F19 gene
          Gene Aliases:
          1700020I02Rik, BB065030
          Chromosome Location:
          Chr.14: 76677568 - 76687108 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01315246_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700108F19Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015485.1 4 - 5 594 71
          GenBank mRNA AK006165.1 - 3 - 4 1154 71
          AK007133.1 - 4 - 5 584 71
          BY707214.1 - 4 - 5 582 71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CB271207.1 - 4 - 5 550 Chr. 14 - 76677612 - 76686890 [+] on Build GRCm38 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream stress-associated endoplasmic reticulum protein family member 2 Serp2 2810032E02Rik -122.2kb Anti-sense Chr. 14 - 76532812 - 76557919 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930431P22 gene 4930431P22Rik 52.7kb Sense Chr. 14 - 76739843 - 76763266 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001074.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2051179 AK076702.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 328647 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144341 - -
          Whole Mouse Genome G4122A A_52_P574247 AK133110.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574247 NR_015485.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574247 AK006165.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574247 AK007133.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P574247 AK076702.1 mouse_transcriptome
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11734504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11443527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11784521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11259060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11259061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11438650 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735294 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11257156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11443530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11443533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29735290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11259055 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1430202_PM_at - -
          Mouse Genome 430 2.0 Array 1430202_at - -
          Mouse Genome 430 2.0 Array 1430202_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400008644.mm.1 TC1400001074.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400008644.mm.1 NR_015485.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400008644.mm.1 NR_015485.1 Refseq
          Murine Genome U74v2 Set 168744_at - -
          Murine Genome U74v2 Set 168744_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430202_at - -
          Affymetrix HT Mouse Genome 430B Array 1430202_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 931898 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0