Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01316188_m1
          See other FGFR1OP GE Assays ›
          Gene Symbol:
          Fgfr1op
          Gene Name:
          Fgfr1 oncogene partner
          Gene Aliases:
          4930553O10Rik, FOP
          Chromosome Location:
          Chr.17: 8165485 - 8196477 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01316188_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgfr1op
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001197046.1 NP_001183975.1 10 - 11 1072 106
          NM_201230.5 NP_957682.1 9 - 10 1012 106
          GenBank mRNA AK016110.1 - 10 - 11 1075 106
          AK028778.1 - 9 - 10 941 106
          AK031261.1 - 9 - 10 995 106
          AK050389.1 - 9 - 10 950 106
          BC050774.1 - 9 - 10 964 106
          BC058804.1 - 9 - 10 947 106
          BC080717.1 - 10 - 11 1059 4935625 106
          BC108331.1 - 10 - 11 1070 30434232 106

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000081.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgfr1op

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein kinase activity
          positive regulation of cell population proliferation
          positive regulation of cell growth
          positive regulation of cell migration
          microtubule anchoring
          negative regulation of protein tyrosine kinase activity
          protein kinase binding
          protein tyrosine kinase inhibitor activity
          protein homodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700000632.mm.1 TC1700000081.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000632.mm.1 NM_001197046.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000632.mm.1 NM_001197046.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000632.mm.1 NM_201230.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000632.mm.1 NM_201230.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000634.mm.1 TC1700000081.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000634.mm.1 NM_001197046.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000634.mm.1 NM_001197046.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000634.mm.1 NM_201230.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000634.mm.1 NM_201230.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000633.mm.1 TC1700000081.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700000633.mm.1 NM_001197046.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000633.mm.1 NM_001197046.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000633.mm.1 NM_201230.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700000633.mm.1 NM_201230.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-85f78c8899-kf4dt:80/100.66.130.62:80.
          git-commit: 7f1fbef35c8161921da6e3861d81379c0ca55d8f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix