Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01316282_g1
          See other SLC39A2 GE Assays ›
          Gene Symbol:
          Slc39a2
          Gene Name:
          solute carrier family 39 (zinc transporter), member 2
          Gene Aliases:
          F730005G13Rik, Gm1789, Gm289, Zip2
          Chromosome Location:
          Chr.14: 51891972 - 51896745 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01316282_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc39a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039676.2 NP_001034765.2 3 - 4 487 98
          GenBank mRNA AK089322.1 - 3 - 4 439 98
          AK170178.1 - 3 - 4 442 98
          BC113769.1 - 3 - 4 457 40057862 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002799.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          zinc ion transport
          zinc ion transmembrane transport
          zinc ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343733 BC113769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343733 AK089322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343733 NM_001039676.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343733 BC113769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343733 AK089322.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P343733 NM_001039676.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 706355 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647662 AK047988.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 23068 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343733 BC113769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343733 AK089322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343733 NM_001039676.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343733 BC113769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343733 AK089322.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P343733 NM_001039676.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603616 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08111642 - -
          Whole Mouse Genome G4122A A_51_P343733 BC113769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343733 AK089322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343733 NM_001039676.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343733 BC113769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343733 AK089322.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P343733 NM_001039676.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08111642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11777125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11777118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11268649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11777114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11777111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11777120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11268643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603640 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11245230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11268644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11777119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11777131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11245229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603634 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10414613 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10414615 - -
          Affymetrix HT MG-430 PM Array Plate 1443163_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10414612 - -
          Mouse Genome 430 2.0 Array 1443163_PM_at - -
          Mouse Genome 430 2.0 Array 1443163_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214922_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214922_at - -
          Affymetrix Mouse Clariom S Array PSR1400004691.mm.1 TC1400002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004691.mm.1 NM_001039676.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004691.mm.1 NM_001039676.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004689.mm.1 TC1400002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004689.mm.1 NM_001039676.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004689.mm.1 NM_001039676.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004688.mm.1 TC1400002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004688.mm.1 NM_001039676.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004688.mm.1 NM_001039676.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004687.mm.1 TC1400002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004687.mm.1 NM_001039676.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004687.mm.1 NM_001039676.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004692.mm.1 TC1400002799.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400004692.mm.1 NM_001039676.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400004692.mm.1 NM_001039676.2 Refseq
          Mouse Expression Set 430 1443163_PM_at - -
          Mouse Expression Set 430 1443163_at - -
          Affymetrix HT Mouse Genome 430B Array 1443163_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443163_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 455644 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2696656 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2510187 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2696656 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0