Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01316380_m1
          See other DDHD1 GE Assays ›
          Gene Symbol:
          Ddhd1
          Gene Name:
          DDHD domain containing 1
          Gene Aliases:
          Mir5131, PA-PLA1
          Chromosome Location:
          Chr.14: 45593171 - 45658143 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01316380_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddhd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039106.3 NP_001034195.2 10 - 11 2332 78
          NM_001042719.2 NP_001036184.1 11 - 12 2434 78
          NM_001284399.1 NP_001271328.1 12 - 13 2406 78
          NM_176845.5 NP_789815.3 10 - 11 2332 78
          GenBank mRNA AK014975.1 - 8 - 9 1118 78
          AK048690.1 - 11 - 12 1892 78
          AK129425.1 - 8 - 9 1743 78
          AK155294.1 - 12 - 13 2363 78
          AK170271.1 - 11 - 12 2046 78
          BC018552.1 - 2 - 3 379 78
          BC043475.1 - 9 - 10 1586 5365658 78
          CA315511.1 - 1 - 2 105 78
          DQ272481.1 - 10 - 11 2331 78
          DQ272482.1 - 10 - 11 2341 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001906.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddhd1

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          lipid catabolic process
          sperm mitochondrion organization
          positive regulation of mitochondrial fission
          molecular_function
          phospholipase activity
          prenylcysteine methylesterase activity
          lipase activity
          hydrolase activity
          1-oxa-2-oxocycloheptane lactonase activity
          sulfolactone hydrolase activity
          3,4-dihydrocoumarin hydrolase activity
          butyrolactone hydrolase activity
          endosulfan lactone lactonase activity
          L-ascorbate 6-phosphate lactonase activity
          Ser-tRNA(Thr) hydrolase activity
          Ala-tRNA(Pro) hydrolase activity
          Cys-tRNA(Pro) hydrolase activity
          Ser(Gly)-tRNA(Ala) hydrolase activity
          metal ion binding
          all-trans-retinyl-palmitate hydrolase, all-trans-retinol forming activity
          retinyl-palmitate esterase activity
          mannosyl-oligosaccharide 1,6-alpha-mannosidase activity
          mannosyl-oligosaccharide 1,3-alpha-mannosidase activity
          methyl indole-3-acetate esterase activity
          methyl salicylate esterase activity
          methyl jasmonate esterase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400014415.mm.1 TC1400001906.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400014415.mm.1 NM_001039106.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014415.mm.1 NM_001042719.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014415.mm.1 NM_001284399.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014415.mm.1 NM_176845.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014416.mm.1 TC1400001906.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400014416.mm.1 NM_001039106.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014416.mm.1 NM_001042719.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014416.mm.1 NM_001284399.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400014416.mm.1 NM_176845.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0