Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300022033.mm.1 |
TC0300002800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022033.mm.1 |
NM_023214.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022033.mm.1 |
NM_023214.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022030.mm.1 |
TC0300002800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022030.mm.1 |
NM_023214.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022030.mm.1 |
NM_023214.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022032.mm.1 |
TC0300002800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022032.mm.1 |
NM_023214.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022032.mm.1 |
NM_023214.7 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022031.mm.1 |
TC0300002800.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300022031.mm.1 |
NM_023214.7.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300022031.mm.1 |
NM_023214.7 |
Refseq |