Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01317892_m1
          See other BLM GE Assays ›
          Gene Symbol:
          Blm
          Gene Name:
          Bloom syndrome, RecQ helicase-like
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 80454993 - 80535119 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01317892_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Blm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042527.2 NP_001035992.1 15 - 16 3122 126
          NM_007550.4 NP_031576.4 15 - 16 3393 126
          GenBank mRNA AB008674.1 - 15 - 16 3383 126
          AK137022.1 - 15 - 16 3105 126
          Z98263.1 - 15 - 16 3100 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004655.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Blm

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cyclin-dependent protein serine/threonine kinase activity
          telomere maintenance
          DNA double-strand break processing
          DNA replication
          DNA repair
          DNA recombination
          cellular response to DNA damage stimulus
          mitotic G2 DNA damage checkpoint signaling
          response to X-ray
          replication fork processing
          DNA duplex unwinding
          cellular metabolic process
          positive regulation of transcription, DNA-templated
          negative regulation of DNA recombination
          negative regulation of mitotic recombination
          alpha-beta T cell differentiation
          positive regulation of alpha-beta T cell proliferation
          regulation of binding
          protein complex oligomerization
          chromosome organization
          negative regulation of cell division
          negative regulation of thymocyte apoptotic process
          cellular response to ionizing radiation
          cellular response to hydroxyurea
          cellular response to camptothecin
          nucleotide binding
          bubble DNA binding
          p53 binding
          nucleic acid binding
          DNA binding
          DNA helicase activity
          single-stranded DNA binding
          catalytic activity
          helicase activity
          protein binding
          ATP binding
          four-way junction helicase activity
          hydrolase activity
          hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
          ATP hydrolysis activity
          ATP-dependent DNA/DNA annealing activity
          3'-5' DNA helicase activity
          G-quadruplex DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700030269.mm.1 TC0700004655.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700030269.mm.1 NM_001042527.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030269.mm.1 NM_001042527.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030269.mm.1 NM_007550.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700030269.mm.1 NM_007550.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0