Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01318741_m1
          See other HPRT GE Assays ›
          Gene Symbol:
          Hprt
          Gene Name:
          hypoxanthine guanine phosphoribosyl transferase
          Gene Aliases:
          C81579, HPGRT, Hprt1
          Chromosome Location:
          Chr.X: 52988078 - 53021660 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01318741_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hprt
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013556.2 NP_038584.2 1 - 2 169 148
          GenBank mRNA AK002286.1 - 1 - 2 114 148
          AK082827.1 - 1 - 2 59 148
          AK088114.1 - 1 - 2 59 148
          AK146626.1 - 1 - 2 115 148
          BC004686.1 - 1 - 2 84 3500885 148
          BC083145.1 - 1 - 2 55 30445781 148
          J00423.1 - 1 - 2 110 148

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000471.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK002286.1, AK088114.1, AK146626.1, BC004686.1, BC083145.1, J00423.1, NM_013556.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hprt

          Gene Ontology Categories:

          Function(s) Process(es)

          response to amphetamine
          purine nucleotide biosynthetic process
          purine ribonucleoside salvage
          adenine salvage
          guanine salvage
          grooming behavior
          locomotory behavior
          nucleoside metabolic process
          cytolysis
          striatum development
          cerebral cortex neuron differentiation
          central nervous system neuron development
          GMP salvage
          IMP salvage
          dopamine metabolic process
          hypoxanthine salvage
          positive regulation of dopamine metabolic process
          GMP catabolic process
          IMP metabolic process
          adenine metabolic process
          hypoxanthine metabolic process
          lymphocyte proliferation
          dendrite morphogenesis
          protein homotetramerization
          nucleotide binding
          magnesium ion binding
          hypoxanthine phosphoribosyltransferase activity
          transferase activity
          glycosyltransferase activity
          identical protein binding
          protein homodimerization activity
          metal ion binding
          guanine phosphoribosyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00003094.mm.1 TC0X00000471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003094.mm.1 NM_013556.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003094.mm.1 NM_013556.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003092.mm.1 TC0X00000471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003092.mm.1 NM_013556.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003092.mm.1 NM_013556.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003093.mm.1 TC0X00000471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003093.mm.1 NM_013556.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003093.mm.1 NM_013556.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003091.mm.1 TC0X00000471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003091.mm.1 NM_013556.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003091.mm.1 NM_013556.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003095.mm.1 TC0X00000471.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003095.mm.1 NM_013556.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003095.mm.1 NM_013556.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0