Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01318833_mH
          See other CLCA3B GE Assays ›
          Gene Symbol:
          Clca3b
          Gene Name:
          chloride channel calcium activated 3B
          Gene Aliases:
          Clca4
          Chromosome Location:
          Chr.3: 144822623 - 144849302 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01318833_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clca3b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_139148.2 NP_631887.2 13 - 14 2393 63
          GenBank mRNA AY008277.1 - 13 - 14 2393 63
          BC045599.1 - 1 - 2 64 63
          BC058766.1 - 13 - 14 2375 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003080.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clca3b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          chloride transmembrane transport
          molecular_function
          intracellular calcium activated chloride channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158176 BC058766.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158176 AY008277.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158176 BC045599.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158176 NM_139148.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 630121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808471 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613204 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 556981 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546162 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 237373 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 4347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 267858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 163170 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71513 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45605 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129012 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144286 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158176 BC058766.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158176 AY008277.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158176 BC045599.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158176 NM_139148.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P507602 BC058766.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P507602 AY008277.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P507602 BC045599.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P507602 NM_139148.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605319 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605331 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605343 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22605340 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129012 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05105277 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05105254 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144286 - -
          Whole Mouse Genome G4122A A_51_P507602 BC058766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P507602 AY008277.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P507602 BC045599.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P507602 NM_139148.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144286 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129012 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03184432 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03184424 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03184428 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03184430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22605347 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02978607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03150090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02669299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02290208 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02669292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605299 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605281 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03102310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02669294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22605329 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10502580 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502581 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502586 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502585 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502577 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502582 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502587 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502576 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502579 - -
          Affymetrix Mouse Gene 1.0 ST Array 10502583 - -
          Affymetrix HT MG-430 PM Array Plate 1451823_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10502575 - -
          Mouse Genome 430 2.0 Array 1451823_PM_at - -
          Mouse Genome 430 2.0 Array 1451823_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 229927_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 229927_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300024210.mm.1 TC0300003080.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024210.mm.1 NM_139148.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024210.mm.1 NM_139148.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024207.mm.1 TC0300003080.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024207.mm.1 NM_139148.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024207.mm.1 NM_139148.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024209.mm.1 TC0300003080.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024209.mm.1 NM_139148.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024209.mm.1 NM_139148.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024208.mm.1 TC0300003080.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300024208.mm.1 NM_139148.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300024208.mm.1 NM_139148.2 Refseq
          Mouse Expression Set 430 1451823_PM_at - -
          Mouse Expression Set 430 1451823_at - -
          Mouse Genome 430A 2.0 Array 1451823_PM_at - -
          Mouse Genome 430A 2.0 Array 1451823_at - -
          Affymetrix HT Mouse Genome 430A Array 1451823_at - -
          Affymetrix HT Mouse Genome 430A Array 1451823_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 380204 - -
          ABI Mouse Genome Survey Microarray 595781 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 70072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1257159 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257159 - -
          mouse-6 v1.1 (Illumina) scl00229927.1_282-S - -
          Illumina Sentrix mouse-6.v1 bead array 70072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00229927.1_282-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1257159 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257159 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0