Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01318966_m1
          See other MMP14 GE Assays ›
          Gene Symbol:
          Mmp14
          Gene Name:
          matrix metallopeptidase 14 (membrane-inserted)
          Gene Aliases:
          AI325305, MT-MMP-1, MT1-MMP, sabe
          Chromosome Location:
          Chr.14: 54431604 - 54441258 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01318966_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mmp14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008608.3 NP_032634.3 5 - 6 1108 82
          GenBank mRNA AK040968.1 - 1081 82
          AK088476.1 - 5 - 6 1108 82
          AK134340.1 - 5 - 6 1039 82
          AK138611.1 - 5 - 6 1049 82
          AK149636.1 - 5 - 6 1037 82
          AK149907.1 - 5 - 6 1088 82
          AK149988.1 - 5 - 6 1008 82
          AK155117.1 - 5 - 6 1110 82
          AK163601.1 - 5 - 6 1100 82
          AK165014.1 - 5 - 6 1109 82
          AK180818.1 - 1 - 2 85 82
          AK212086.1 - 1 - 2 59 82
          BC076638.1 - 5 - 6 1100 30658833 82
          DQ249870.1 - 5 - 6 846 82
          U54984.1 - 5 - 6 1092 82
          X83536.2 - 5 - 6 1018 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000745.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mmp14

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          endochondral ossification
          regulation of transcription, DNA-templated
          proteolysis
          response to hormone
          positive regulation of peptidase activity
          cell migration
          positive regulation of cell growth
          lung development
          positive regulation of cell migration
          collagen catabolic process
          zymogen activation
          chondrocyte proliferation
          embryonic cranial skeleton morphogenesis
          branching morphogenesis of an epithelial tube
          tissue remodeling
          negative regulation of focal adhesion assembly
          bone development
          craniofacial suture morphogenesis
          DNA-binding transcription factor activity
          metalloendopeptidase activity
          integrin binding
          calcium ion binding
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          peptidase activator activity
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400005815.mm.1 TC1400000745.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400005815.mm.1 NM_008608.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-66756dbf47-27kf9:80/100.66.130.62:80.
          git-commit: ed967532dff6834307dac93af002e5c8e22ba7c9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline