Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01319186_m1
          See other DLEU2 GE Assays ›
          Gene Symbol:
          Dleu2
          Gene Name:
          deleted in lymphocytic leukemia, 2
          Gene Aliases:
          1810047A16Rik, 4932411A06Rik, 7730401J12Rik, AI197277, AI788824, Alt1, Gm9069, Leu2
          Chromosome Location:
          Chr.14: 61602836 - 61682373 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01319186_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dleu2
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028264.1
          1 - 2 264 87
          GenBank mRNA AF380423.1 - LIT3422
          DLEU2
          1 - 2 241 87
          AK042416.1 -
          1 - 2 217 87
          AK080165.1 -
          1 - 2 67 87
          AK084358.1 -
          1 - 2 220 87
          AK134888.1 -
          1 - 2 206 87
          AK165901.1 -
          1 - 2 254 87
          BB639368.1 -
          1 - 2 264 87
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY329982.1 - 1 - 2 74 Chr. 14 - 61625311 - 61682182 [-] on Build GRCm38 87
          GenBankmRNA BY773229.1 - 1 - 2 254 Chr. 14 - 61635552 - 61682362 [-] on Build GRCm38 87
          GenBankmRNA AI789372.1 - 1 - 2 200 Chr. 14 - 61603658 - 61682309 [-] on Build GRCm38 87
          GenBankmRNA AI047357.1 - 1 - 2 218 Chr. 14 - 61603621 - 61682327 [-] on Build GRCm38 87
          GenBankmRNA BY727464.1 - 1 - 2 206 Chr. 14 - 61635227 - 61682314 [-] on Build GRCm38 87
          GenBankmRNA BB658096.1 - 1 - 2 216 Chr. 14 - 61603603 - 61682325 [-] on Build GRCm38 87
          GenBankmRNA BY102972.1 - 1 - 2 223 Chr. 14 - 61635556 - 61682325 [-] on Build GRCm38 87
          GenBankmRNA BI697003.1 - 1 - 2 196 Chr. 14 - 61603472 - 61682304 [-] on Build GRCm38 87
          GenBankmRNA BY182555.1 - 1 - 2 220 Chr. 14 - 61635532 - 61682327 [-] on Build GRCm38 87
          GenBankmRNA BB639334.1 - 1 - 2 217 Chr. 14 - 61635523 - 61682325 [-] on Build GRCm38 87
          GenBankmRNA BY718835.1 - 1 - 2 206 Chr. 14 - 61614174 - 61682314 [-] on Build GRCm38 87
          GenBankmRNA BY775788.1 - 1 - 2 254 Chr. 14 - 61643355 - 61682362 [-] on Build GRCm38 87
          GenBankmRNA BY734056.1 - 1 - 2 254 Chr. 14 - 61603574 - 61682362 [-] on Build GRCm38 87
          GenBankmRNA AA171261.1 - 1 - 2 204 Chr. 14 - 61603747 - 61682316 [-] on Build GRCm38 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          27017
          Gm27017 Anti-sense Chr. 14 - 61680039 - 61686642 on NCBI Build GRCm38
          Downstream deleted in lymphocytic leukemia,
          7
          Dleu7 BC038059 593.9kb Sense Chr. 14 - 62276229 - 62292979 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400002195.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of neuron projection development
          negative regulation of neuron maturation
          molecular_function

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1400017015.mm.1 TC1400002195.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400017015.mm.1 NR_028264.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017015.mm.1 NR_028264.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017016.mm.1 TC1400002195.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400017016.mm.1 NR_028264.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017016.mm.1 NR_028264.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017027.mm.1 TC1400002195.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400017027.mm.1 NR_028264.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017027.mm.1 NR_028264.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017031.mm.1 TC1400002195.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400017031.mm.1 NR_028264.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400017031.mm.1 NR_028264.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 921581 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0