Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01319804_g1
          See other GM29678 GE Assays ›
          Gene Symbol:
          Gm29678
          Gene Name:
          predicted gene, 29678
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 190403648 - 190417006 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01319804_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm29678
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK085057.1 - 2 - 3 234 66

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC102632463 LOC102632463 -198.7kb Sense Chr. 1 - 190170295 - 190217279 on NCBI Build GRCm38
          Downstream ribosomal protein S6 kinase polypeptide 1 Rps6kc1 AA682037,
          B130003F20Rik,
          C80612,
          Rpk118
          355.7kb Anti-sense Chr. 1 - 190772879 - 190912139 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444709 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04398345 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01142705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00736147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00354289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00354293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00029461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01142691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01076131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932145 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00354286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00715155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00318933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00223170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01142588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00354292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932235 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00373867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20932168 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 102900672 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244783 - -
          Illumina Sentrix mouse-6.v1 bead array 102900672 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102900672 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244783 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0