Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01320199_m1
          See other GM16897 GE Assays ›
          Gene Symbol:
          Gm16897
          Gene Name:
          predicted gene, 16897
          Gene Aliases:
          C030002c11rik
          Chromosome Location:
          Chr.1: 194955759 - 194976959 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01320199_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm16897
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033564.1 4 - 5 1472 111
          GenBank mRNA AK030377.1 - 4 - 5 1473 111

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 3962 Mir3962 mmu-mir-3962,
          mu-mir-3962
          -133.7kb Anti-sense Chr. 1 - 194822001 - 194822064 on NCBI Build GRCm38
          Downstream RIKEN cDNA A330023F24 gene A330023F24Rik C030002C11Rik 58.2kb Anti-sense Chr. 1 - 195017399 - 195037908 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100003850.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 155492 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847139 BC066020.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847139 NM_001005788.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 97119 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 167466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 953288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464061 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156731 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137099 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143371 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143371 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00290472 - -
          Whole Mouse Genome G4122A A_52_P471340 AK030377.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P471340 NR_033564.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201139 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958800 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00230307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00640471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00294274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00201121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20958788 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100031507.mm.1 TC0100003850.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100031507.mm.1 NR_033564.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100031507.mm.1 NR_033564.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 877583 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0