Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400008903.mm.1 |
TC1400001112.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008906.mm.1 |
TC1400001112.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008906.mm.1 |
NM_001030294.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008906.mm.1 |
NM_001030294.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008907.mm.1 |
TC1400001112.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008907.mm.1 |
NM_001030294.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008907.mm.1 |
NM_001030294.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008905.mm.1 |
TC1400001112.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008905.mm.1 |
NM_001030294.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008905.mm.1 |
NM_001030294.1 |
Refseq |