Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022985.6 | NP_075361.2 | 1 - 2 | 41 | 69 | |
GenBank mRNA | AK002775.1 | - | 1 - 2 | 42 | 69 |
Set Membership: |
![]() ![]() ![]() ![]() |