Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01323289_m1
          See other CCDC160 GE Assays ›
          Gene Symbol:
          Ccdc160
          Gene Name:
          coiled-coil domain containing 160
          Gene Aliases:
          6330534C20Rik
          Chromosome Location:
          Chr.X: 52791200 - 52799474 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01323289_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc160
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034059.1 NP_001029231.1 1 - 2 127 103
          GenBank mRNA AK134708.1 - 1 - 2 127 103
          BC138946.1 - 1 - 2 75 8861968 103
          BC138947.1 - 1 - 2 75 8861969 103

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000466.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK134708.1, BC138946.1, BC138947.1, NM_001034059.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32384238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32384241 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32384239 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32384240 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092399 NM_001034059.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2092399 AK134708.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115594 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452915 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734486 AK030059.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 578600 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139070 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092399 NM_001034059.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2092399 AK134708.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09293707 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09293693 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09293707 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32384239 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32384238 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32384241 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32384240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32384207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14811608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14584180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14681205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14681206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14700152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15163505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14579634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14579639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15192554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14590455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15095489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15066225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14811609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15163521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14784637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14700153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14811604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384281 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32384344 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10599606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10599605 - -
          Affymetrix HT MG-430 PM Array Plate 1437936_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10599604 - -
          Mouse Genome 430 2.0 Array 1437936_PM_at - -
          Mouse Genome 430 2.0 Array 1437936_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 434778_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 434778_at - -
          Affymetrix Mouse Clariom S Array PSR0X00003063.mm.1 TC0X00000466.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003063.mm.1 NM_001034059.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003063.mm.1 NM_001034059.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003061.mm.1 TC0X00000466.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003061.mm.1 NM_001034059.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003061.mm.1 NM_001034059.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003060.mm.1 TC0X00000466.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003060.mm.1 NM_001034059.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003060.mm.1 NM_001034059.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003062.mm.1 TC0X00000466.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00003062.mm.1 NM_001034059.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00003062.mm.1 NM_001034059.1 Refseq
          Mouse Expression Set 430 1437936_at - -
          Mouse Expression Set 430 1437936_PM_at - -
          Murine Genome U74v2 Set 131915_PM_at - -
          Murine Genome U74v2 Set 131915_at - -
          Affymetrix HT Mouse Genome 430B Array 1437936_at - -
          Affymetrix HT Mouse Genome 430B Array 1437936_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 332828 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3161818 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3161818 - -
          mouse-6 v1.1 (Illumina) GI_85702180-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3161819 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3161819 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0