Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01323704_m1
          See other 4930554H23RIK GE Assays ›
          Gene Symbol:
          4930554H23Rik
          Gene Name:
          RIKEN cDNA 4930554H23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 63344682 - 63444562 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01323704_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930554H23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK019764.1 - 9 - 10 2176 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cholinergic receptor,
          nicotinic,
          alpha polypeptide 7
          Chrna7 Acra7,
          alpha7,
          nAchR
          -132.2kb Sense Chr. 7 - 63098692 - 63212526 on NCBI Build GRCm38
          Downstream OTU domain containing 7A Otud7a AJ430384,
          Otud7
          96.7kb Anti-sense Chr. 7 - 63444745 - 63759905 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2311780 AK019764.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 433530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931113 AK050042.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931113 AK050325.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931113 AK154151.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931113 BC024105.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931113 NM_022026.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511461 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 288400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877356 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445839 AK041899.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445839 AK042434.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445839 BC114544.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445839 D38581.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445839 NM_010695.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223085 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133705 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23474 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537233 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722617 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415166 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 815049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462967 AK077804.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 860871 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26173 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 161608 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 925736 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222030 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 130790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 522049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636969 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 195469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 313795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822920 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 148481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447427 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 18214 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143709 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67822 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 84599 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139587 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06216914 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01530305 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06216909 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06216991 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139587 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06216922 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01530254 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01530284 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125733 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01530284 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139587 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06216922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25171785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25171769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25171771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06602189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25171780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25171775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06602187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25171777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25171768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25171781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25171774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25171778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25171783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06451923 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454020_PM_at - -
          Mouse Genome 430 2.0 Array 1454020_PM_at - -
          Mouse Genome 430 2.0 Array 1454020_at - -
          Mouse Expression Set 430 1454020_PM_at - -
          Mouse Expression Set 430 1454020_at - -
          Affymetrix HT Mouse Genome 430B Array 1454020_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454020_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0