Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01324451_g1
          See other CIZ1 GE Assays ›
          Gene Symbol:
          Ciz1
          Gene Name:
          CDKN1A interacting zinc finger protein 1
          Gene Aliases:
          0610038H21Rik, 2900056O04Rik, LSFR1, ZNF356
          Chromosome Location:
          Chr.2: 32363006 - 32378313 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01324451_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ciz1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252534.1 NP_001239463.1 1 - 2 457 171
          NM_001252536.1 NP_001239465.1 2 - 3 181 171
          NM_001252537.1 NP_001239466.1 1 - 2 457 171
          GenBank mRNA AK089986.1 - 1 - 2 460 171
          AK159103.1 - 1 - 2 435 171
          AK165687.1 - 2 - 3 135 171
          BC072624.1 - 302 6825309 171

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000577.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK089986.1, AK159103.1, AK165687.1, BC072624.1, NM_001252534.1, NM_001252536.1, NM_001252537.1 transcripts.

          Set Membership:

          Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of DNA-dependent DNA replication initiation
          maintenance of protein location in nucleus
          cyclin binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200005151.mm.1 TC0200000577.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005151.mm.1 NM_001252534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005151.mm.1 NM_001252534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005151.mm.1 NM_001252537.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005151.mm.1 NM_001252537.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005146.mm.1 TC0200000577.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005146.mm.1 NM_001252536.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005146.mm.1 NM_001252536.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005146.mm.1 NM_001252538.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005146.mm.1 NM_001252538.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005153.mm.1 TC0200000577.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005153.mm.1 NM_001252534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005153.mm.1 NM_001252534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005153.mm.1 NM_001252537.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005153.mm.1 NM_001252537.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005154.mm.1 TC0200000577.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005154.mm.1 NM_001252534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005154.mm.1 NM_001252534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005154.mm.1 NM_001252536.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005154.mm.1 NM_001252536.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005154.mm.1 NM_001252537.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005154.mm.1 NM_001252537.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005145.mm.1 TC0200000577.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005145.mm.1 NM_001252536.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005145.mm.1 NM_001252536.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005145.mm.1 NM_001252538.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005145.mm.1 NM_001252538.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005150.mm.1 TC0200000577.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005150.mm.1 NM_001252534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005150.mm.1 NM_001252534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005150.mm.1 NM_001252537.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005150.mm.1 NM_001252537.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005152.mm.1 TC0200000577.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005152.mm.1 NM_001252534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005152.mm.1 NM_001252534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005152.mm.1 NM_001252537.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005152.mm.1 NM_001252537.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 590220 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0