Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01324729_mH
          See other PFKP GE Assays ›
          Gene Symbol:
          Pfkp
          Gene Name:
          phosphofructokinase, platelet
          Gene Aliases:
          1200015H23Rik, 9330125N24Rik, ATP-PFK, PFK-C, PFK-P
          Chromosome Location:
          Chr.13: 6579874 - 6648789 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01324729_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pfkp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001291071.1 NP_001278000.1 5 - 6 713 96
          NM_019703.4 NP_062677.1 5 - 6 713 96
          GenBank mRNA AF123533.1 - 5 - 6 678 96
          AK004790.1 - 5 - 6 668 96
          AK020367.1 - 5 - 6 668 96
          AK049841.1 - 5 - 6 667 96
          AK076803.1 - 5 - 6 668 96
          AK078254.1 - 5 - 6 668 96
          AK098162.1 - 5 - 6 667 96
          AK152670.1 - 5 - 6 668 96
          AK154125.1 - 5 - 6 668 96
          AK165422.1 - 5 - 6 667 96
          AK165425.1 - 5 - 6 667 96
          AK170624.1 - 5 - 6 668 96
          AK171062.1 - 5 - 6 668 96
          AY779275.1 - 5 - 6 630 96
          AY779276.1 - 5 - 6 630 96
          BC006926.1 - 5 - 6 652 2655383 96
          CT010268.1 - 5 - 6 621 96
          Y19008.1 - 5 - 6 670 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001395.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pfkp

          Gene Ontology Categories:

          Function(s) Process(es)

          fructose 6-phosphate metabolic process
          glycolytic process
          metabolic process
          phosphorylation
          fructose 1,6-bisphosphate metabolic process
          carbohydrate phosphorylation
          protein homotetramerization
          positive regulation of mitophagy in response to mitochondrial depolarization
          nucleotide binding
          catalytic activity
          6-phosphofructokinase activity
          ATP binding
          kinase activity
          transferase activity
          protein-containing complex binding
          metal ion binding
          fructose-6-phosphate binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1300008765.mm.1 TC1300001395.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008765.mm.1 NM_019703.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008766.mm.1 TC1300001395.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008766.mm.1 NM_019703.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0