Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01325287_g1
          See other CTC1 GE Assays ›
          Gene Symbol:
          Ctc1
          Gene Name:
          CTS telomere maintenance complex component 1
          Gene Aliases:
          1500010J02Rik, AAF-132, AAF132
          Chromosome Location:
          Chr.11: 69015904 - 69036473 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01325287_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013256.2 NP_001013274.2 18 - 19 3217 78
          NM_001143790.1 NP_001137262.1 18 - 19 3214 78
          NM_001281464.1 NP_001268393.1 18 - 19 3085 78
          NM_001281465.1 NP_001268394.1 18 - 19 3201 78
          GenBank mRNA AK038651.1 - 18 - 19 3001 78
          AK044055.1 - 18 - 19 2996 78
          AK145919.1 - 18 - 19 3032 78
          AK165906.1 - 18 - 19 3017 78
          AK186509.1 - 2 - 3 265 78
          AK214175.1 - 3 - 4 337 78
          AK215862.1 - 1 - 2 52 78
          BC014687.1 - 9 - 10 1385 78
          BC060629.1 - 14 - 15 2267 6847619 78
          BC066078.1 - 18 - 19 3028 78
          BC072625.1 - 18 - 19 3042 78
          BC147495.1 - 18 - 19 3015 9056325 78
          BC147497.1 - 18 - 19 3015 9056354 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000891.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctc1

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance
          cellular response to DNA damage stimulus
          aging
          regulation of G2/M transition of mitotic cell cycle
          telomere maintenance via telomere lengthening
          multicellular organism growth
          positive regulation of DNA replication
          positive regulation of fibroblast proliferation
          spleen development
          thymus development
          bone marrow development
          chromosome organization
          hematopoietic stem cell proliferation
          replicative senescence
          DNA binding
          single-stranded DNA binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100007468.mm.1 TC1100000891.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100007468.mm.1 NM_001013256.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007468.mm.1 NM_001013256.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007468.mm.1 NM_001143790.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007468.mm.1 NM_001143790.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007468.mm.1 NM_001281464.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100007468.mm.1 NM_001281465.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0