Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aif1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145144.1 | NP_660126.1 | 1 - 2 | 75 | 60 | |
GenBank mRNA | AB035322.1 | - | 1 - 2 | 27 | 60 | |
AK028955.1 | - | 1 - 2 | 33 | 60 | ||
AK045539.1 | - | 1 - 2 | 105 | 60 | ||
BC024599.1 | - | 1 - 2 | 75 | 5044495 | 60 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |