Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01327403_g1
          See other SLC27A4 GE Assays ›
          Gene Symbol:
          Slc27a4
          Gene Name:
          solute carrier family 27 (fatty acid transporter), member 4
          Gene Aliases:
          BB144259, FATP4
          Chromosome Location:
          Chr.2: 29802680 - 29817522 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01327403_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc27a4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_011989.4 NP_036119.1 10 - 11 1750 98
          GenBank mRNA AF072759.1 - 8 - 9 1052 98
          AJ251113.1 - 9 - 10 1463 98
          AK009292.1 - 4 - 5 537 98
          AK033519.1 - 6 - 7 694 98
          AK036919.1 - 10 - 11 1752 98
          AK155313.1 - 10 - 11 1719 98
          AK155388.1 - 10 - 11 1720 98
          AK170377.1 - 10 - 11 1754 98
          AK215383.1 - 2 - 3 206 98
          BC023114.1 - 10 - 11 1601 4158956 98
          CT010312.1 - 9 - 10 1463 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000511.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc27a4

          Gene Ontology Categories:

          Function(s) Process(es)

          very long-chain fatty acid metabolic process
          medium-chain fatty acid transport
          long-chain fatty acid metabolic process
          lipid metabolic process
          fatty acid metabolic process
          transport
          lipid transport
          metabolic process
          fatty acid transport
          long-chain fatty acid transport
          very long-chain fatty acid catabolic process
          skin development
          long-chain fatty acid import into cell
          nucleotide binding
          catalytic activity
          long-chain fatty acid-CoA ligase activity
          succinate-CoA ligase activity
          3-oxo-2-(2'-pentenyl)cyclopentane-1-octanoic acid CoA ligase activity
          fatty acid ligase activity
          ligase activity
          3-isopropenyl-6-oxoheptanoyl-CoA synthetase activity
          2-oxo-delta3-4,5,5-trimethylcyclopentenylacetyl-CoA synthetase activity
          benzoyl acetate-CoA ligase activity
          2,4-dichlorobenzoate-CoA ligase activity
          very long-chain fatty acid-CoA ligase activity
          pivalate-CoA ligase activity
          cyclopropanecarboxylate-CoA ligase activity
          adipate-CoA ligase activity
          citronellyl-CoA ligase activity
          mentha-1,3-dione-CoA ligase activity
          thiophene-2-carboxylate-CoA ligase activity
          2,4,4-trimethylpentanoate-CoA ligase activity
          cis-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
          trans-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
          acetate-CoA ligase (ADP-forming) activity
          methylbutanoate-CoA ligase activity
          3-hydroxypropionyl-CoA synthetase activity
          perillic acid-CoA ligase (ADP-forming) activity
          perillic acid-CoA ligase (AMP-forming) activity
          (3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (ADP-forming) activity
          (3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (AMP-forming) activity
          pristanate-CoA ligase activity
          malonyl-CoA synthetase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200003764.mm.1 TC0200000511.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200003764.mm.1 NM_011989.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200003764.mm.1 NM_011989.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0