Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01327827_m1
          See other PTPRJ GE Assays ›
          Gene Symbol:
          Ptprj
          Gene Name:
          protein tyrosine phosphatase, receptor type, J
          Gene Aliases:
          AI450271, BET, Byp, CD148, DEP-1, PTPbeta2, Ptpb2, RPTPJ, Scc-1, Scc1
          Chromosome Location:
          Chr.2: 90429756 - 90580677 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01327827_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptprj
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001135657.1 NP_001129129.1 7 - 8 1428 78
          NM_008982.5 NP_033008.3 7 - 8 1712 78
          GenBank mRNA AK147318.1 - 7 - 8 1633 78
          AK147556.1 - 7 - 8 1633 78
          AY038891.1 - 7 - 8 1530 78
          D45212.1 - 7 - 8 1668 78
          DQ133576.1 - 7 - 8 1443 78

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptprj

          Gene Ontology Categories:

          Function(s) Process(es)

          vasculogenesis
          positive regulation of cell-matrix adhesion
          protein dephosphorylation
          heart development
          blood coagulation
          negative regulation of cell population proliferation
          positive regulation of platelet activation
          negative regulation of platelet-derived growth factor receptor signaling pathway
          positive regulation of macrophage chemotaxis
          dephosphorylation
          regulation of cell adhesion
          negative regulation of cell growth
          negative regulation of cell migration
          peptidyl-tyrosine dephosphorylation
          negative regulation of epidermal growth factor receptor signaling pathway
          negative regulation of vascular permeability
          negative regulation of MAP kinase activity
          positive regulation of cell adhesion
          platelet-derived growth factor receptor signaling pathway
          oligodendrocyte differentiation
          negative regulation of T cell receptor signaling pathway
          positive chemotaxis
          positive regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          negative regulation of protein kinase B signaling
          negative regulation of signaling receptor activity
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          platelet-derived growth factor receptor binding
          beta-catenin binding
          hydrolase activity
          phosphatase activity
          protein kinase binding
          gamma-catenin binding
          mitogen-activated protein kinase binding
          delta-catenin binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-85f78c8899-kf4dt:80/100.66.130.62:80.
          git-commit: 7f1fbef35c8161921da6e3861d81379c0ca55d8f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix