Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01327926_g1
          See other MEG3 GE Assays ›
          Gene Symbol:
          Meg3
          Gene Name:
          maternally expressed 3
          Gene Aliases:
          2900016C05Rik, 3110050O07Rik, 6330408G06Rik, AI425946, AW108224, D12Bwg1266e, Gtl2, R74756, R75394
          Chromosome Location:
          Chr.12: 109540996 - 109571729 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01327926_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Meg3
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003633.3
          2 - 3 1219 82
          NR_027651.2
          6 - 7 1207 82
          NR_027652.1
          7 - 8 1332 82
          GenBank mRNA AK018105.1 -
          2 - 3 1220 82
          AK141629.1 -
          2 - 3 132 82
          AK147321.1 -
          4 - 5 2687 82
          BC024818.1 -
          4 - 5 579 82
          BC051667.1 -
          496 82
          BC094421.1 -
          6 - 7 1196 82
          Y13832.1 - LIT3453
          Gtl2
          7 - 8 1323 82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA396833.1 - 213 Chr. - - [-] on Build 81
          GenBankmRNA CB194103.1 - 2 - 3 157 Chr. 12 - 109549289 - 109571721 [+] on Build GRCm38 82
          GenBankmRNA CF584712.1 - 536 Chr. - - [-] on Build 82
          GenBankmRNA CF746141.1 - 3 - 4 236 Chr. 12 - 109544694 - 109558925 [+] on Build GRCm38 82
          GenBankmRNA AA183389.1 - 2 - 3 Chr. 12 - 109549269 - 109558284 [+] on Build GRCm38 80
          GenBankmRNA BM729672.1 - 2 - 3 98 Chr. 12 - 109549348 - 109571555 [+] on Build GRCm38 82
          GenBankmRNA DN174347.1 - 4 - 5 349 Chr. 12 - 109544662 - 109568602 [+] on Build GRCm38 82
          GenBankmRNA AA004059.1 - 2 - 3 Chr. 12 - 109546470 - 109549810 [+] on Build GRCm38 79
          GenBankmRNA BF120932.1 - 4 - 5 355 Chr. 12 - 109544664 - 109571540 [+] on Build GRCm38 82
          GenBankmRNA CD776077.1 - 2 - 3 Chr. 12 - 109549476 - 109571726 [-] on Build GRCm38 82
          GenBankmRNA BF658505.1 - 2 - 3 191 Chr. 12 - 109549255 - 109571463 [+] on Build GRCm38 82
          GenBankmRNA AV584558.1 - 2 - 3 212 Chr. 12 - 109549234 - 109571423 [+] on Build GRCm38 82
          GenBankmRNA CF578092.1 - 3 - 4 Chr. 12 - 109549171 - 109571726 [-] on Build GRCm38 82
          GenBankmRNA CB059150.1 - 2 - 3 226 Chr. 12 - 109549228 - 109571595 [+] on Build GRCm38 82
          GenBankmRNA CF585122.1 - 3 - 4 398 Chr. 12 - 109546306 - 109571648 [+] on Build GRCm38 82
          GenBankmRNA BI965904.1 - 3 - 4 342 Chr. 12 - 109546357 - 109549793 [+] on Build GRCm38 82
          GenBankmRNA W64614.1 - 147 Chr. - - [-] on Build 81
          GenBankmRNA BM894630.1 - 2 - 3 158 Chr. 12 - 109549288 - 109571555 [+] on Build GRCm38 82
          GenBankmRNA BI965842.1 - 3 - 4 358 Chr. 12 - 109546340 - 109549809 [+] on Build GRCm38 82
          GenBankmRNA CO042160.1 - 3 - 4 Chr. 12 - 109549291 - 109571726 [-] on Build GRCm38 82
          GenBankmRNA AW742476.1 - 3 - 4 322 Chr. 12 - 109544608 - 109558301 [+] on Build GRCm38 82
          GenBankmRNA BP775209.1 - 1 - 2 Chr. 12 - 109544722 - 109549805 [-] on Build GRCm38 82
          GenBankmRNA BM053819.1 - 3 - 4 342 Chr. 12 - 109546356 - 109549803 [+] on Build GRCm38 82
          GenBankmRNA BF471019.1 - 3 - 4 440 Chr. 12 - 109544490 - 109558300 [+] on Build GRCm38 82
          GenBankmRNA BU058056.1 - 481 Chr. - - [-] on Build 82
          GenBankmRNA CK030896.1 - 4 - 5 437 Chr. 12 - 109544574 - 109571455 [+] on Build GRCm38 82
          GenBankmRNA BQ954402.1 - 1 - 2 95 Chr. 12 - 109549476 - 109559105 [+] on Build GRCm38 82
          GenBankmRNA AA103529.1 - 1 - 2 Chr. 12 - 109549491 - 109571625 [+] on Build GRCm38 79
          GenBankmRNA CB202480.1 - 375 Chr. - - [-] on Build 82
          GenBankmRNA CF584711.1 - 3 - 4 Chr. 12 - 109549157 - 109571726 [-] on Build GRCm38 82
          GenBankmRNA AV583825.1 - 4 - 5 323 Chr. 12 - 109544985 - 109558255 [+] on Build GRCm38 82
          GenBankmRNA CF584777.1 - 517 Chr. - - [-] on Build 82
          GenBankmRNA CB590891.1 - 158 Chr. - - [-] on Build 82
          GenBankmRNA AV480686.1 - 2 - 3 232 Chr. 12 - 109549224 - 109558320 [+] on Build GRCm38 82
          GenBankmRNA CF745386.1 - 1 - 2 39 Chr. 12 - 109549514 - 109559125 [+] on Build GRCm38 82
          GenBankmRNA BY718623.1 - 2 - 3 223 Chr. 12 - 109546394 - 109558814 [+] on Build GRCm38 82
          GenBankmRNA BM053951.1 - 3 - 4 342 Chr. 12 - 109546356 - 109549790 [+] on Build GRCm38 82
          GenBankmRNA AV502014.1 - 2 - 3 166 Chr. 12 - 109549280 - 109558307 [+] on Build GRCm38 82
          GenBankmRNA BE855234.1 - 2 - 3 257 Chr. 12 - 109549189 - 109558320 [+] on Build GRCm38 82
          GenBankmRNA EL605878.1 - 2 - 3 Chr. 12 - 109546466 - 109558320 [-] on Build GRCm38 82
          GenBankmRNA CF618073.1 - 2 - 3 313 Chr. 12 - 109549157 - 109571607 [+] on Build GRCm38 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 1906-1 Mir1906-1 Mir1906,
          Mirn1906,
          mmu-mir-1906,
          mmu-mir-1906-1
          -4.9kb Anti-sense Chr. 12 - 109544541 - 109544620 on NCBI Build GRCm38
          Downstream microRNA 770 Mir770 Mirn770,
          mir-770,
          mmu-mir-770
          13.9kb Anti-sense Chr. 12 - 109563692 - 109563785 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200001127.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s)

          in utero embryonic development
          liver development
          DNA methylation
          skeletal muscle tissue development
          determination of adult lifespan
          post-embryonic development
          multicellular organism growth
          lung alveolus development
          SMAD protein signal transduction
          genetic imprinting

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200008057.mm.1 TC1200001127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008057.mm.1 NR_003633.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008057.mm.1 NR_027651.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008057.mm.1 NR_027652.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008057.mm.1 NR_027652.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008060.mm.1 TC1200001127.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200008060.mm.1 NR_003633.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008060.mm.1 NR_027651.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008060.mm.1 NR_027652.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200008060.mm.1 NR_027652.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0