Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01328473_m1
          See other BRSK1 GE Assays ›
          Gene Symbol:
          Brsk1
          Gene Name:
          BR serine/threonine kinase 1
          Gene Aliases:
          Gm1100, SAD-B, SADB
          Chromosome Location:
          Chr.7: 4690928 - 4715997 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01328473_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Brsk1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001003920.3 NP_001003920.2 17 - 18 2242 81
          GenBank mRNA AY533671.1 - 17 - 18 2084 81
          BC086636.1 - 17 - 18 2241 6402113 81
          BC086803.1 - 4 - 5 608 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000038.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Brsk1

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          protein phosphorylation
          cellular response to DNA damage stimulus
          cell cycle
          neurotransmitter secretion
          nervous system development
          axonogenesis
          response to UV
          phosphorylation
          peptidyl-serine phosphorylation
          establishment of cell polarity
          neuron differentiation
          intracellular signal transduction
          neuron projection morphogenesis
          centrosome duplication
          nucleotide binding
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein kinase binding
          gamma-tubulin binding
          metal ion binding
          tau-protein kinase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700000496.mm.1 TC0700000038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000496.mm.1 NM_001003920.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000496.mm.1 NM_001003920.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000498.mm.1 TC0700000038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000498.mm.1 NM_001003920.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000498.mm.1 NM_001003920.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000495.mm.1 TC0700000038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000495.mm.1 NM_001003920.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000495.mm.1 NM_001003920.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000497.mm.1 TC0700000038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000497.mm.1 NM_001003920.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000497.mm.1 NM_001003920.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000499.mm.1 TC0700000038.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700000499.mm.1 NM_001003920.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700000499.mm.1 NM_001003920.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0