Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01328599_g1
          See other MALSU1 GE Assays ›
          Gene Symbol:
          Malsu1
          Gene Name:
          mitochondrial assembly of ribosomal large subunit 1
          Gene Aliases:
          2410003K15Rik, Matsu1
          Chromosome Location:
          Chr.6: 49073795 - 49139144 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01328599_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Malsu1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029353.1 NP_083629.1 3 - 4 570 61
          GenBank mRNA AK010366.1 - 3 - 4 512 61
          AK031878.1 - 3 - 4 521 61
          AK153503.1 - 2 - 3 339 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000578.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of translation
          ribosomal large subunit biogenesis
          negative regulation of mitochondrial translation
          negative regulation of ribosome biogenesis
          molecular_function
          ribosomal large subunit binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311096 AK010366.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311096 AK031878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311096 AK153503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311096 AV085631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311096 BC022611.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311096 AK010366.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311096 AK031878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311096 AK153503.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311096 AV085631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P311096 BC022611.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 507639 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 851782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698205 AK041088.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698205 BC107232.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698205 BC107233.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698205 NM_177115.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 82733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131584 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33025 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146125 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145061 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311096 AK010366.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311096 AK031878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311096 AK153503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311096 AV085631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311096 BC022611.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311096 AK010366.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311096 AK031878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311096 AK153503.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311096 AV085631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P311096 BC022611.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311096 AK010366.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311096 AK031878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311096 AK153503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311096 AV085631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311096 BC022611.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311096 AK010366.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311096 AK031878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311096 AK153503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311096 AV085631.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P311096 BC022611.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343853 AK163220.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343853 BC049082.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343853 AK088465.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343853 AK147140.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P343853 NM_023670.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P324748 AK010366.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P324748 AK031878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P324748 AK153503.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P324748 BC022611.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P324748 NM_029353.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05122940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24430818 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1428495_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10538201 - -
          Mouse Genome 430 2.0 Array 1459016_at - -
          Mouse Genome 430 2.0 Array 1428495_at - -
          Mouse Genome 430 2.0 Array 1459016_PM_at - -
          Mouse Genome 430 2.0 Array 1428495_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600004467.mm.1 TC0600000578.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600004467.mm.1 NM_029353.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600004467.mm.1 NM_029353.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600004468.mm.1 TC0600000578.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600004468.mm.1 NM_029353.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600004468.mm.1 NM_029353.1 Refseq
          Mouse Expression Set 430 1428495_at - -
          Mouse Expression Set 430 1428495_PM_at - -
          Murine Genome U74v2 Set 132381_at - -
          Murine Genome U74v2 Set 132381_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428495_at - -
          Affymetrix HT Mouse Genome 430B Array 1428495_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 510011 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0