Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200031340.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031340.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031340.mm.1 |
NM_018733.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031341.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031341.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031341.mm.1 |
NM_018733.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031343.mm.1 |
TC0200003674.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031343.mm.1 |
NM_018733.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031343.mm.1 |
NM_018733.2 |
Refseq |