Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01329917_g1
          See other IFI27L2B GE Assays ›
          Gene Symbol:
          Ifi27l2b
          Gene Name:
          interferon, alpha-inducible protein 27 like 2B
          Gene Aliases:
          1810023F06Rik, ISG12(b2)
          Chromosome Location:
          Chr.12: 103450898 - 103457223 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01329917_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifi27l2b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145449.2 NP_663424.1 6 - 7 638 78
          GenBank mRNA AK007590.1 - 4 - 5 529 78
          BC021795.1 - 6 - 7 638 4217067 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200002270.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifi27l2b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085779 NM_145449.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085779 AK007590.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085779 BC021795.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085776 NM_145449.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085776 BC021795.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864658 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339466 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 735551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 235940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 10770 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153877 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085776 NM_145449.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085776 BC021795.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085779 NM_145449.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085779 AK007590.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085779 BC021795.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28697697 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153877 - -
          Whole Mouse Genome G4122A A_51_P391432 NM_145449.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391432 AK007590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P391432 BC021795.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153877 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10340654 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09780065 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10340658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28697659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10171617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10340651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10340658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28697675 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10402357 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402356 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402358 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402359 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402354 - -
          Affymetrix Mouse Gene 1.0 ST Array 10402355 - -
          Affymetrix HT MG-430 PM Array Plate 1425120_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10402353 - -
          Mouse Genome 430 2.0 Array 1425120_PM_x_at - -
          Mouse Genome 430 2.0 Array 1425120_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217845_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217845_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1200015764.mm.1 TC1200002270.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200015764.mm.1 NM_145449.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200015764.mm.1 NM_145449.2 Refseq
          Mouse Expression Set 430 1425120_x_at - -
          Mouse Expression Set 430 1425120_PM_x_at - -
          Murine Genome U74v2 Set 168934_at - -
          Murine Genome U74v2 Set 165944_PM_at - -
          Murine Genome U74v2 Set 168934_PM_at - -
          Murine Genome U74v2 Set 165944_at - -
          Mouse Genome 430A 2.0 Array 1425120_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1425120_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425120_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1425120_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 644676 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5570048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232995 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232995 - -
          mouse-6 v1.1 (Illumina) scl42110.7.200_45-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl42110.7.200_45-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232995 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232995 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0