Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01330358_m1
          See other CAPN3 GE Assays ›
          Gene Symbol:
          Capn3
          Gene Name:
          calpain 3
          Gene Aliases:
          AI323605, Capa-3, Capa3, Lp82, p94
          Chromosome Location:
          Chr.2: 120463593 - 120504919 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01330358_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Capn3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109761.2 NP_001103231.1 11 - 12 1406 61
          NM_001177799.1 NP_001171270.1 11 - 12 1673 61
          NM_007601.3 NP_031627.2 12 - 13 1817 61
          GenBank mRNA AF091998.1 - 11 - 12 1370 61
          AF127766.2 - 11 - 12 1393 61
          BC090661.1 - 11 - 12 1406 30681055 61
          BC139790.1 - 12 - 13 1537 40047690 61
          X92523.1 - 12 - 13 1537 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200005480.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Capn3

          Gene Ontology Categories:

          Function(s) Process(es)

          autolysis
          proteolysis
          positive regulation of satellite cell activation involved in skeletal muscle regeneration
          response to muscle activity
          myofibril assembly
          negative regulation of protein sumoylation
          negative regulation of apoptotic process
          regulation of I-kappaB kinase/NF-kappaB signaling
          sarcomere organization
          regulation of myoblast differentiation
          positive regulation of proteolysis
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          regulation of catalytic activity
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of release of sequestered calcium ion into cytosol
          response to calcium ion
          muscle structure development
          protein-containing complex assembly
          G1 to G0 transition involved in cell differentiation
          cellular response to calcium ion
          cellular response to salt stress
          protein localization to membrane
          self proteolysis
          sodium-dependent self proteolysis
          catalytic activity
          calcium-dependent cysteine-type endopeptidase activity
          signaling receptor binding
          calcium ion binding
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          structural constituent of muscle
          hydrolase activity
          enzyme binding
          sodium ion binding
          titin binding
          metal ion binding
          ligase regulator activity
          molecular adaptor activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200013701.mm.1 TC0200005480.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200013701.mm.1 NM_001109761.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013701.mm.1 NM_001109761.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013701.mm.1 NM_001177799.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013701.mm.1 NM_001177799.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013701.mm.1 NM_007601.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200013701.mm.1 NM_007601.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0