Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01331066_m1
          See other ACVR1 GE Assays ›
          Gene Symbol:
          Acvr1
          Gene Name:
          activin A receptor, type 1
          Gene Aliases:
          ALK2, ActR-I, ActRIA, Acvr, Acvrlk2, Alk-2, Alk8, D330013D15Rik, SKR1, Tsk7L
          Chromosome Location:
          Chr.2: 58446438 - 58566828 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01331066_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acvr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110204.1 NP_001103674.1 5 - 6 740 72
          NM_001110205.1 NP_001103675.1 3 - 4 218 72
          NM_007394.3 NP_031420.2 4 - 5 495 72
          GenBank mRNA AF332087.1 - 2 - 3 128 72
          AF332088.1 - 2 - 3 128 72
          AK052246.1 - 330 72
          AK053428.1 - 5 - 6 739 72
          BC058718.1 - 2 - 3 216 6418954 72
          L15436.1 - 3 - 4 568 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003579.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acvr1

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          branching involved in blood vessel morphogenesis
          urogenital system development
          in utero embryonic development
          gastrulation with mouth forming second
          mesoderm formation
          neural crest cell migration
          embryonic heart tube morphogenesis
          mitral valve morphogenesis
          atrial septum primum morphogenesis
          protein phosphorylation
          transmembrane receptor protein serine/threonine kinase signaling pathway
          transforming growth factor beta receptor signaling pathway
          germ cell development
          determination of left/right symmetry
          gastrulation
          mesoderm development
          heart development
          embryo development
          negative regulation of signal transduction
          positive regulation of pathway-restricted SMAD protein phosphorylation
          phosphorylation
          peptidyl-threonine phosphorylation
          regulation of ossification
          positive regulation of bone mineralization
          BMP signaling pathway
          activin receptor signaling pathway
          negative regulation of activin receptor signaling pathway
          intracellular signal transduction
          positive regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          smooth muscle cell differentiation
          pharyngeal system development
          pathway-restricted SMAD protein phosphorylation
          cardiac muscle cell fate commitment
          endocardial cushion cell fate commitment
          positive regulation of determination of dorsal identity
          negative regulation of extrinsic apoptotic signaling pathway
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          transmembrane receptor protein serine/threonine kinase activity
          transforming growth factor beta receptor activity, type I
          ATP binding
          kinase activity
          activin receptor activity, type I
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          growth factor binding
          protein homodimerization activity
          SMAD binding
          metal ion binding
          activin binding
          transforming growth factor beta binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200030482.mm.1 TC0200003579.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200030482.mm.1 NM_001110204.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030482.mm.1 NM_001110204.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030482.mm.1 NM_001110205.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030482.mm.1 NM_001110205.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030482.mm.1 NM_007394.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030482.mm.1 NM_007394.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030484.mm.1 TC0200003579.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200030484.mm.1 NM_001110204.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030484.mm.1 NM_001110204.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030485.mm.1 TC0200003579.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200030485.mm.1 NM_001110204.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030485.mm.1 NM_001110204.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030485.mm.1 NM_007394.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030485.mm.1 NM_007394.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030486.mm.1 TC0200003579.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200030486.mm.1 NM_001110204.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030486.mm.1 NM_001110204.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030486.mm.1 NM_007394.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030486.mm.1 NM_007394.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030487.mm.1 TC0200003579.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200030487.mm.1 NM_001110204.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030487.mm.1 NM_001110204.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030487.mm.1 NM_007394.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030487.mm.1 NM_007394.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0