Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01331449_m1
          See other GM13483 GE Assays ›
          Gene Symbol:
          Gm13483
          Gene Name:
          predicted gene 13483
          Gene Aliases:
          OTTMUSG00000012382
          Chromosome Location:
          Chr.2: 50296810 - 50433967 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01331449_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13483
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040362.1 1 - 2 95 96
          GenBank mRNA AK045825.1 - 1 - 2 41 96
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BB644362.1 - 1 - 2 41 Chr. 2 - 50296865 - 50433981 [+] on Build GRCm38 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream methylmalonic aciduria (cobalamin deficiency) cblD type,
          with homocystinuria
          Mmadhc 2010311D03Rik,
          AI314967
          -66.0 Anti-sense Chr. 2 - 50279881 - 50296799 on NCBI Build GRCm38
          Downstream heat shock protein 90 alpha (cytosolic),
          class B member 1 pseudogene
          Gm13498 OTTMUSG00000012426 476.4kb Sense Chr. 2 - 50909684 - 50911846 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000805.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239769 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175394 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 554653 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670337 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 751884 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50010 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 294816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 960384 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 702192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 143134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74610 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 16099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 496811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 681258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931939 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 217938 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 956409 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926601 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178850 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 640487 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 602089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 47785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 908770 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 733652 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126817 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 559110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351061 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 109529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 493489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 14132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212336 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 178263 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 374861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 823804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666232 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 268720 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 674687 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884820 AK017304.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884820 AK029377.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884820 BC047134.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884820 BC080814.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884820 BC083121.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884820 NM_001025574.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884820 NM_175167.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884820 XM_976257.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 175301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 109727 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 166549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114571 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110175 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 149188 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119327 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00365957 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04514376 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119327 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04514359 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04514503 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04514415 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04514522 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04514452 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04514389 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04514548 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125598 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04514359 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04514415 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04514548 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21208039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21208026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01433375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207505 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21208040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21208035 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207434 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02267040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01914101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02267028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207741 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21208044 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207770 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01278174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207748 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01263056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207507 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207437 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21208032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01168842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207772 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01239737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01603180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21208028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02283000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01168854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01196211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02177837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01778189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01278168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01373536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21208027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01278178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02216654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207464 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207504 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01278171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02177826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21208041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21208037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01762669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21207631 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200006700.mm.1 TC0200000805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006700.mm.1 NR_040361.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006700.mm.1 NR_040361.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006700.mm.1 NR_040362.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006700.mm.1 NR_040362.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006705.mm.1 TC0200000805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006705.mm.1 NR_040362.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006705.mm.1 NR_040362.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006704.mm.1 TC0200000805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006704.mm.1 NR_040362.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006704.mm.1 NR_040362.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006699.mm.1 TC0200000805.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200006699.mm.1 NR_040361.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006699.mm.1 NR_040361.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006699.mm.1 NR_040362.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200006699.mm.1 NR_040362.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 595644 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0