Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01331455_m1
          See other BB557941 GE Assays ›
          Gene Symbol:
          BB557941
          Gene Name:
          expressed sequence BB557941
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 57127477 - 57181754 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01331455_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BB557941
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040356.1 2 - 3 191 74
          GenBank mRNA AK163349.1 - 2 - 3 192 74
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY725773.1 - 2 - 3 192 Chr. 2 - 57127920 - 57181754 [-] on Build GRCm38 74

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream nuclear receptor subfamily 4,
          group A,
          member 2
          Nr4a2 HZF-3,
          NOT,
          Nurr1,
          RNR-1,
          TINOR,
          TINUR
          -3.5kb Sense Chr. 2 - 57106830 - 57124003 on NCBI Build GRCm38
          Downstream RIKEN cDNA 4930555B11 gene 4930555B11Rik 52.8kb Anti-sense Chr. 2 - 57181831 - 57188257 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003562.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2319020 AK163349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2319020 BB557941.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140053 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 744917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961441 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832785 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29179 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 856288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 115228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715667 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461867 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 897202 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386712 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 671337 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348056 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 469825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 99348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 392686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755817 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131118 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79979 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161732 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2319020 AK163349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2319020 BB557941.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04528025 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04528078 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04528102 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00375691 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161732 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240155 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240117 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01771658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02246623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01211631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01348560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240325 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01892795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240147 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01912731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887399 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240257 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01793265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02226828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02166123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01912718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240151 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01793260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01795004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02166130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02022130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01793261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01565377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240141 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240170 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01867571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02246622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01532477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01348562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240131 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01348559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02166115 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02022125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01887401 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01793263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01683262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01383561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02218433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240157 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02085806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02061562 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240113 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21240311 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1443368_PM_at - -
          Mouse Genome 430 2.0 Array 1443368_PM_at - -
          Mouse Genome 430 2.0 Array 1443368_at - -
          Affymetrix Mouse Clariom S Array PSR0200030364.mm.1 TC0200003562.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200030364.mm.1 NR_040356.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200030364.mm.1 NR_040356.1 Refseq
          Mouse Expression Set 430 1443368_PM_at - -
          Mouse Expression Set 430 1443368_at - -
          Affymetrix HT Mouse Genome 430B Array 1443368_at - -
          Affymetrix HT Mouse Genome 430B Array 1443368_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0