Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01331462_m1
          See other 8430437L04RIK GE Assays ›
          Gene Symbol:
          8430437L04Rik
          Gene Name:
          RIKEN cDNA 8430437L04 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 72703510 - 72729468 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01331462_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 8430437L04Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040503.1 6 - 7 1049 115
          GenBank mRNA AK018472.1 - 6 - 7 1049 115

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cell division cycle associated 7 Cdca7 2310021G01Rik,
          AU044109,
          JPO1
          -239.9kb Sense Chr. 2 - 72476189 - 72486892 on NCBI Build GRCm38
          Downstream RIKEN cDNA 6430710C18 gene 6430710C18Rik 5033406J01Rik 16.4kb Anti-sense Chr. 2 - 72745846 - 72813717 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001061.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2289527 AK018472.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 560305 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295703 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 334200 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444130 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252490 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442713 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736319 AK135058.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736319 AK154671.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736319 BC002185.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736319 BC030465.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736319 BC034283.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 736319 NM_029153.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 162170 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153176 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2289527 AK018472.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04563911 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04563957 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110339 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04563957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327335 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02201730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01457338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327339 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01457341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327338 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327345 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01757215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01457340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21327349 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1431978_PM_at - -
          Mouse Genome 430 2.0 Array 1431978_PM_at - -
          Mouse Genome 430 2.0 Array 1431978_at - -
          Affymetrix Mouse Clariom S Array PSR0200008675.mm.1 TC0200001061.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200008675.mm.1 NR_040503.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008675.mm.1 NR_040503.1 Refseq
          Mouse Expression Set 430 1431978_PM_at - -
          Mouse Expression Set 430 1431978_at - -
          Affymetrix HT Mouse Genome 430B Array 1431978_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1431978_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 497286 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101740114 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0