Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01331490_m1
          See other WT1OS GE Assays ›
          Gene Symbol:
          Wt1os
          Gene Name:
          Wilms tumor 1 homolog, opposite strand
          Gene Aliases:
          6030498F17, AI314831, Wt1-as
          Chromosome Location:
          Chr.2: 105076538 - 105126510 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01331490_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wt1os
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015462.1 5 - 6 1125 71
          GenBank mRNA AK031729.1 - 2 - 3 977 71
          AK032905.1 - 2 - 3 446 71
          AK165089.1 - 5 - 6 1127 71
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI526688.1 - 2 - 3 328 Chr. 2 - 105078218 - 105089166 [-] on Build GRCm38 71
          GenBankmRNA BX523040.1 - 2 - 3 352 Chr. 2 - 105079589 - 105089166 [-] on Build GRCm38 71
          GenBankmRNA AI316915.1 - 2 - 3 328 Chr. 2 - 105078251 - 105089166 [-] on Build GRCm38 71
          GenBankmRNA BB623434.1 - 2 - 3 446 Chr. 2 - 105079577 - 105089284 [-] on Build GRCm38 71
          GenBankmRNA AI786893.1 - 2 - 3 275 Chr. 2 - 105078271 - 105089116 [-] on Build GRCm38 71

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream eukaryotic translation initiation factor 3,
          subunit M
          Eif3m Ga17,
          Pcid1,
          Tango7
          -62.6kb Sense Chr. 2 - 104999656 - 105017027 on NCBI Build GRCm38
          Downstream Wilms tumor 1 homolog Wt1 D630046I19Rik,
          Wt-1
          40.7kb Anti-sense Chr. 2 - 105126529 - 105173614 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004257.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK165089.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P555739 AK032905.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P555739 AK165089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P555739 NR_015462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P555739 AK031729.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P555739 AK032905.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P555739 AK165089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P555739 NR_015462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P555739 AK031729.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074661 AK032905.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074661 AK165089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074661 NR_015462.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074661 AK031729.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94870 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 820836 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 240700 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 373682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 642149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517611 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13865 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767985 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 350898 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 314949 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813753 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 844526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 932527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 32266 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750293 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 512248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 953605 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233357 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 594927 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436712 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 707007 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 700550 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 411230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147685 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 56143 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58477 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88497 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136177 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175458 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P555739 AK032905.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P555739 AK165089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P555739 NR_015462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P555739 AK031729.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P555739 AK032905.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P555739 AK165089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P555739 NR_015462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P555739 AK031729.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074661 AK032905.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074661 AK165089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074661 NR_015462.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074661 AK031729.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136177 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00451101 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04633403 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04633439 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175458 - -
          Whole Mouse Genome G4122A A_52_P555739 AK032905.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P555739 AK165089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P555739 NR_015462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P555739 AK031729.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P555739 AK032905.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P555739 AK165089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P555739 NR_015462.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P555739 AK031729.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117806 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136177 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01646936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01646934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492285 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01646933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492332 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01315556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01646925 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01646932 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01646921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01646926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02042122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01646922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01646945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01893875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01606491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01646928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492404 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01646931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21492403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01167878 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10485644 - -
          Affymetrix HT MG-430 PM Array Plate 1457698_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10485643 - -
          Mouse Genome 430 2.0 Array 1457698_at - -
          Mouse Genome 430 2.0 Array 1457698_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200035852.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035852.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035852.mm.1 NR_015462.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035847.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035847.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035847.mm.1 NR_015462.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035855.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035855.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035855.mm.1 NR_015462.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035861.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035861.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035861.mm.1 NR_015462.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035844.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035844.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035844.mm.1 NR_015462.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035850.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035850.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035850.mm.1 NR_015462.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035856.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035856.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035856.mm.1 NR_015462.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035860.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035860.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035860.mm.1 NR_015462.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035846.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035846.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035846.mm.1 NR_015462.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035849.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035849.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035849.mm.1 NR_015462.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035848.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035848.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035848.mm.1 NR_015462.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035845.mm.1 TC0200004257.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035845.mm.1 NR_015462.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035845.mm.1 NR_015462.1 Refseq
          Mouse Expression Set 430 1457698_PM_at - -
          Mouse Expression Set 430 1457698_at - -
          Affymetrix HT Mouse Genome 430B Array 1457698_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457698_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 608626 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105570110 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0