Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01332254_m1
          See other PAK3 GE Assays ›
          Gene Symbol:
          Pak3
          Gene Name:
          p21 protein (Cdc42/Rac)-activated kinase 3
          Gene Aliases:
          PAK-3, Pak65alpha, Pak65beta, Stk4
          Chromosome Location:
          Chr.X: 143518368 - 143797796 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01332254_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pak3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001195046.1 NP_001181975.1 14 - 15 1540 97
          NM_001195047.1 NP_001181976.1 13 - 14 1495 97
          NM_001195048.1 NP_001181977.1 14 - 15 1650 97
          NM_001195049.1 NP_001181978.1 16 - 17 1914 97
          NM_008778.3 NP_032804.2 13 - 14 1523 97
          GenBank mRNA AF082297.1 - 12 - 13 1437 97
          AJ496262.1 - 12 - 13 1405 97
          AJ496263.1 - 13 - 14 1450 97
          AK031853.1 - 14 - 15 1531 97
          AK158893.1 - 14 - 15 1650 97
          AM943853.1 - 13 - 14 1468 97
          AM943854.1 - 14 - 15 1513 97
          BC053403.1 - 13 - 14 1530 30060082 97
          U39738.1 - 17 - 18 1921 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00001413.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pak3

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein phosphorylation
          Rho protein signal transduction
          multicellular organism development
          regulation of mitotic cell cycle
          axonogenesis
          metabolic process
          regulation of neuron projection development
          phosphorylation
          dendrite development
          cell migration
          actin cytoskeleton organization
          regulation of actin filament polymerization
          stress-activated protein kinase signaling cascade
          activation of protein kinase activity
          regulation of apoptotic process
          regulation of MAPK cascade
          positive regulation of neuron apoptotic process
          dendritic spine development
          positive regulation of dendritic spine morphogenesis
          cellular response to organic cyclic compound
          positive regulation of DNA biosynthetic process
          nucleotide binding
          catalytic activity
          protein kinase activity
          protein serine/threonine kinase activity
          MAP kinase kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          SH3 domain binding
          small GTPase binding
          metal ion binding
          GTPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00008977.mm.1 TC0X00001413.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00008977.mm.1 NM_001195046.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008977.mm.1 NM_001195046.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008977.mm.1 NM_001195047.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008977.mm.1 NM_001195047.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008977.mm.1 NM_001195048.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008977.mm.1 NM_001195048.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008977.mm.1 NM_001195049.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008977.mm.1 NM_001195049.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008977.mm.1 NM_008778.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00008977.mm.1 NM_008778.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline