Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01332736_s1
          See other PHXR1 GE Assays ›
          Gene Symbol:
          Phxr1
          Gene Name:
          per-hexamer repeat gene 1
          Gene Aliases:
          SP34
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01332736_s1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Phxr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA X12809.1 - 1 349 124

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962350 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962347 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20962345 - -
          Whole Mouse Genome G4122A A_51_P232275 X12809.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135087 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10468971 - -
          Affymetrix Mouse Gene 1.0 ST Array 10468973 - -
          Affymetrix Mouse Gene 1.0 ST Array 10468972 - -
          Affymetrix HT MG-430 PM Array Plate 1421800_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10468970 - -
          Mouse Genome 430 2.0 Array 1421800_at - -
          Mouse Genome 430 2.0 Array 1421800_PM_at - -
          Mouse Expression Set 430 1421800_at - -
          Mouse Expression Set 430 1421800_PM_at - -
          Murine Genome U74v2 Set 101705_PM_at - -
          Murine Genome U74v2 Set 101705_at - -
          Mouse Genome 430A 2.0 Array 1421800_at - -
          Mouse Genome 430A 2.0 Array 1421800_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421800_at - -
          Affymetrix HT Mouse Genome 430A Array 1421800_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5550041 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005550041 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2757187 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2757187 - -
          mouse-6 v1.1 (Illumina) scl21340.1.1_227-S - -
          Illumina Sentrix mouse-6.v1 bead array 5550041 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21340.1.1_227-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5550041 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2757180 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-865d7754c7-mcq8j:80/100.66.130.62:80.
          git-commit: 7ff36f61b459eb8f02ba5a263f0e3ce367ca82d3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0