Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01333333_mH
          See other DAB2IP GE Assays ›
          Gene Symbol:
          Dab2ip
          Gene Name:
          disabled 2 interacting protein
          Gene Aliases:
          2310011D08Rik, AI480459, Aip1, mKIAA1743
          Chromosome Location:
          Chr.2: 35558458 - 35730994 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01333333_mH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dab2ip
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001001602.2 NP_001001602.2 2 - 3 574 86
          NM_001114124.2 NP_001107596.1 4 - 5 698 86
          NM_001114125.1 NP_001107597.1 4 - 5 550 86
          NM_001290637.1 NP_001277566.1 4 - 5 605 86
          NM_001290639.1 NP_001277568.1 2 - 3 233 86
          NM_001290640.1 NP_001277569.1 2 - 3 538 86
          NM_001290641.1 NP_001277570.1 4 - 5 831 86
          GenBank mRNA AK147464.1 - 4 - 5 630 86
          AK147593.1 - 4 - 5 832 86
          AK155296.1 - 4 - 5 606 86
          AK164475.1 - 4 - 5 613 86
          AY305656.1 - 2 - 3 291 86
          AY305657.1 - 2 - 3 574 86
          AY305658.1 - 2 - 3 233 86
          BC118530.1 - 2 - 3 199 40098202 86
          DQ473307.1 - 4 - 5 435 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000641.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AY305656.1, AY305657.1, AY305658.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dab2ip

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          angiogenesis
          negative regulation of protein phosphorylation
          immune system process
          apoptotic process
          inflammatory response
          response to unfolded protein
          cell cycle
          signal transduction
          I-kappaB phosphorylation
          multicellular organism development
          negative regulation of cell population proliferation
          extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of endothelial cell migration
          negative regulation of epithelial cell migration
          negative regulation of epithelial to mesenchymal transition
          positive regulation of neuron projection development
          negative regulation of phosphatidylinositol 3-kinase signaling
          negative regulation of angiogenesis
          cell motility involved in cerebral cortex radial glia guided migration
          layer formation in cerebral cortex
          negative regulation of cell growth
          negative regulation of vascular endothelial growth factor receptor signaling pathway
          negative regulation of NF-kappaB transcription factor activity
          negative regulation of toll-like receptor 4 signaling pathway
          negative regulation of GTPase activity
          tube formation
          cellular response to vascular endothelial growth factor stimulus
          vascular endothelial growth factor receptor-2 signaling pathway
          reelin-mediated signaling pathway
          regulation of growth
          negative regulation of epidermal growth factor receptor signaling pathway
          positive regulation of apoptotic process
          regulation of GTPase activity
          regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of protein-containing complex assembly
          negative regulation of MAP kinase activity
          positive regulation of MAPK cascade
          regulation of protein heterodimerization activity
          positive regulation of JUN kinase activity
          positive regulation of GTPase activity
          negative regulation of phosphatidylinositol 3-kinase activity
          cellular protein catabolic process
          innate immune response
          positive regulation of protein catabolic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          positive regulation of JNK cascade
          negative regulation of Ras protein signal transduction
          negative regulation of fibroblast proliferation
          neuron projection morphogenesis
          negative regulation of epithelial cell proliferation
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          negative regulation of G0 to G1 transition
          negative regulation of ERK1 and ERK2 cascade
          cellular response to lipopolysaccharide
          cellular response to interleukin-1
          cellular response to tumor necrosis factor
          cellular response to epidermal growth factor stimulus
          negative regulation of protein serine/threonine kinase activity
          positive regulation of protein serine/threonine kinase activity
          negative regulation of canonical Wnt signaling pathway
          positive regulation of synapse maturation
          positive regulation of dendrite development
          regulation of p38MAPK cascade
          negative regulation of vascular endothelial growth factor signaling pathway
          positive regulation of proteasomal protein catabolic process
          negative regulation of cellular protein catabolic process
          positive regulation of neuron migration
          positive regulation of apoptotic signaling pathway
          GTPase activator activity
          death receptor binding
          protein binding
          SH3 domain binding
          kinase binding
          protein kinase binding
          mitogen-activated protein kinase kinase binding
          mitogen-activated protein kinase kinase kinase binding
          phosphatidylinositol-3-phosphate binding
          signaling adaptor activity
          Toll-like receptor 4 binding
          phosphatidylinositol 3-kinase regulatory subunit binding
          identical protein binding
          protein homodimerization activity
          vascular endothelial growth factor receptor 2 binding
          phosphatidylinositol 3-kinase binding
          protein-containing complex binding
          protein N-terminus binding
          protein phosphatase 2A binding
          phosphatidylinositol-4-phosphate binding
          14-3-3 protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200005877.mm.1 TC0200000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005877.mm.1 NM_001001602.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005877.mm.1 NM_001001602.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005884.mm.1 TC0200000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005884.mm.1 NM_001001602.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005884.mm.1 NM_001001602.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005884.mm.1 NM_001114124.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005884.mm.1 NM_001114125.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005884.mm.1 NM_001114125.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005878.mm.1 TC0200000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005878.mm.1 NM_001001602.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005878.mm.1 NM_001001602.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005879.mm.1 TC0200000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005879.mm.1 NM_001001602.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005879.mm.1 NM_001001602.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005883.mm.1 TC0200000641.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200005883.mm.1 NM_001001602.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005883.mm.1 NM_001001602.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005883.mm.1 NM_001114124.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005883.mm.1 NM_001114125.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200005883.mm.1 NM_001114125.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0